Skip to Content
Merck
All Photos(1)

Documents

47775

Supelco

Menadione (K3)

analytical standard

Synonym(s):

Menadione, 2-Methyl-1,4-naphthoquinone, Vitamin K3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O2
CAS Number:
Molecular Weight:
172.18
Beilstein:
1908453
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

feature

standard type fat soluble vitamin

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

105-107 °C (lit.)

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-30°C

SMILES string

CC1=CC(=O)c2ccccc2C1=O

InChI

1S/C11H8O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6H,1H3

InChI key

MJVAVZPDRWSRRC-UHFFFAOYSA-N

Gene Information

human ... NQO1(1728)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Menadione (K3) is a fat-soluble vitamin precursor and catabolic product of oral phylloquinone (vitamin k1) in the intestine.
Menadione is a prothrombogenic compound and belongs to the Vitamin K class of compounds, which are necessary for the biosynthesis of prothrombin and other blood clotting factors. It is used as a model quinone in cell culture and in vivo investigations. It is obtained as a catabolic product of phylloquinone and circulating precursor of tissue menaquinone-4 in rats.

Application

Menadione (K3) may be used as an analytical reference standard for the quantification of the analyte in biological fluids using liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a validated liquid chromatography method for the simultaneous determination of eight fat-soluble vitamins in biological fluids after solid-phase extraction
Chatzimichalakis FP, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 805(2), 289-296 (2004)
Daiki Habu et al.
JAMA, 292(3), 358-361 (2004-07-22)
Previous findings indicate that vitamin K2 (menaquinone) may play a role in controlling cell growth. To determine whether vitamin K2 has preventive effects on the development of hepatocellular carcinoma in women with viral cirrhosis of the liver. Forty women diagnosed
H H Thijssen et al.
The British journal of nutrition, 75(1), 121-127 (1996-01-01)
We measured the vitamin K status in postmortem human tissues (brain, heart, kidney, liver, lung, pancreas) to see if there is a tissue-specific distribution pattern. Phylloquinone (K1) was recovered in all tissues with relatively high levels in liver, heart and
Kerry Routenberg Love et al.
Nature chemical biology, 3(11), 697-705 (2007-10-20)
The addition of ubiquitin (Ub) and ubiquitin-like (Ubl) modifiers to proteins serves to modulate function and is a key step in protein degradation, epigenetic modification and intracellular localization. Deubiquitinating enzymes and Ubl-specific proteases, the proteins responsible for the removal of
Arne Søraas et al.
Journal of oral microbiology, 6 (2014-09-11)
The prevalence of infections caused by Cefotaximase-Munich (CTX-M)-type extended-spectrum beta-lactamase-producing Enterobacteriaceae (ESBL-E) has rapidly increased during the past 15 years. Enterobacteriaceae are commonly found in the gastrointestinal tract and long-term intestinal carriage is considered important for the spread of ESBL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service