Skip to Content
Merck
All Photos(1)

Documents

47358-U

Supelco

EPA 524.2 Fortification Solution

certified reference material, 2000 μg/mL each component in methanol

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 524.1,524.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type fortification

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 524.2 Fortification Solution is a multi-component solution containing 4-bromofluorobenzene, fluorobenzene, and 1,2-dichlorobenzene-d4 as certified reference materials (CRMs).
US EPA 500 Series methods are analytical methods for identifying and quantifying organic contaminants such as volatile organic compounds (VOCs), pesticides, synthetic organic compounds (SOCs), and trihalomethane disinfection byproducts (THMs) in raw source water, finished drinking water, and drinking water at all stages of treatment.

Application

EPA 524.2 Fortification Solution has been used as an internal standard to identify and quantify volatile organic compound (VOC) species of benzene, toluene, ethyltoluene, and xylene (BTEX), bromodichloromethane (BDCM), and THMs, respectively, in river water, tap water samples as well as air and water of indoor swimming pools, by gas chromatography (GC) based techniques. EPA 524.2 Fortification Solution may be used as a CRM to quantify volatile organic compound (VOC) analytes in indoor and outdoor air samples by GC coupled to mass spectrometry (MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    4-Bromofluorobenzene
    1,2-Dichlorobenzene-d4
    Fluorobenzene

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Forensic investigation of BTEX contamination in the Houjing River in Southern Taiwan
Lin C, et al.
Journal of Environmental Engineering and Management, 17(6), 395-395 (2007)
Assessment of air and water contamination by disinfection by-products at 41 indoor swimming pools
Tardif R, et al.
Environmental Research, 148(6), 411-420 (2016)
Low concentrations of bromodichloromethane induce a toxicogenomic response in porcine embryos in vitro
Page-Lariviere F, et al.
Reproductive Toxicology, 66(6), 44-55 (2016)
Volatile organic compounds (VOCs) in conventional and high performance school buildings in the US
Zhong L, et al.
International Journal of Environmental Research and Public Health, 14(1), 100-100 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service