Skip to Content
Merck
All Photos(1)

Key Documents

442747-U

Supelco

Perfluorotributylamine (PFTBA)

analytical standard

Synonym(s):

Heptacosafluorotributylamine, PFTBA, Perfluorotributylamine, Tris(nonafluorobutyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N(CF2CF2CF2CF3)3
CAS Number:
Molecular Weight:
671.09
Beilstein:
1813883
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Agency

EPA 625,8240

vapor density

23.3 (vs air)

vapor pressure

1.3 mmHg ( 25 °C)

CofA

current certificate can be downloaded

feature

standard type tuning solution

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

178 °C (lit.)

density

1.883 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C12F27N/c13-1(14,7(25,26)27)4(19,20)10(34,35)40(11(36,37)5(21,22)2(15,16)8(28,29)30)12(38,39)6(23,24)3(17,18)9(31,32)33

InChI key

RVZRBWKZFJCCIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Perfluorotributylamine (PFTBA) is a neat reference standard, which may be used in environmental analysis.

Application

Perfluorotributylamine (PFTBA) is commonly used for the calibration of the mass axes of mass spectrometers, basically in those systems, which use electron ionization (EI). PFTBA may be used as a calibration standard for the gas chromatography-mass spectrometric determination of tramadol in human plasma samples and phthalate esters in water samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmaceutical Analysis E-Book: A Textbook for Pharmacy Students and Pharmaceutical Chemists, 4 (2015)
Comparison of different fibers for the solid-phase microextraction of phthalate esters from water
Penalver A, et al.
Journal of Chromatography A, 922(1-2), 377-384 (2001)
Validation of a GC/MS method for the determination of tramadol in human plasma after intravenous bolus
Gambaro V, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 58(8), 947-950 (2003)
H A Sudhalkar et al.
Retina (Philadelphia, Pa.), 18(5), 460-465 (1998-11-04)
Perfluorocarbon liquids (PFCLs) are useful for floating various materials off the posterior retina during vitreoretinal surgery. It is generally assumed that buoyancy forces restrict this use to objects with specific gravity lower than that of PFCLs. We tested potential intraocular
N Miki et al.
Journal of orthopaedic science : official journal of the Japanese Orthopaedic Association, 4(5), 370-375 (1999-10-29)
We studied the effects of denervation on the energy metabolism and peripheral circulation dynamics of rat hindlimb muscles during and after exercise. The sciatic nerves of male Wistar rats were cut to produce denervation. Energy metabolism was assessed by phosphorus-31

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service