Skip to Content
Merck
All Photos(1)

Key Documents

40124

Supelco

4,4′-DDT solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane solution, 4,4′-DDT solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9Cl5
CAS Number:
Molecular Weight:
354.49
Beilstein:
1882657
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C(c2ccc(Cl)cc2)C(Cl)(Cl)Cl

InChI

1S/C14H9Cl5/c15-11-5-1-9(2-6-11)13(14(17,18)19)10-3-7-12(16)8-4-10/h1-8,13H

InChI key

YVGGHNCTFXOJCH-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingxiang Zhou et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 48(3), 191-197 (2013-01-30)
The goal of the present study was to investigate the feasibility of silicon dioxide (SiO(2)) microspheres without special modification to enrich dichlorodiphenyltrichloroethane (DDT) and its main metabolites, p,p'-dichlorodiphenyl-2,2-dichloroethylene (p,p'-DDD) and p,p'-dichlorodiphenyldichloroethylene (DDE) in combination with gas chromatography-electron-capture detection. The experimental results
Katarzyna Kulma et al.
PloS one, 8(3), e58322-e58322 (2013-03-14)
Insecticide-resistance threatens the control of mosquito-borne diseases like malaria or dengue fever. To ensure sustainable vector control we need a full understanding of the factors driving the evolution of resistance. We test the hypothesis that the expression of insecticide-resistance depends
Alexandra J White et al.
Environmental health : a global access science source, 12, 24-24 (2013-03-19)
Few studies have supported an association between breast cancer and DDT, usually assessed with biomarkers that cannot discern timing of exposure, or differentiate between the accumulation of chronic low-dose versus acute high-dose exposures in the past. Previous studies suggest that
Maria N Manaca et al.
Environmental science and pollution research international, 20(5), 3472-3479 (2012-11-28)
The concentrations of dichlorodiphenyltrichloroethane (DDT) compounds in cord blood of 214 children born between 2003 and 2006 in Manhiça (Mozambique) have been determined. In this time interval, corresponding to the period before DDT reintroduction for indoor residual spraying, the observed
Shüné V Oliver et al.
Malaria journal, 12, 44-44 (2013-02-02)
Anopheles arabiensis is a major malaria vector in Africa. It thrives in agricultural areas and has been associated with increased malaria incidence in areas under rice and maize cultivation. This effect may be due to increased adult size and abundance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service