Skip to Content
Merck
All Photos(1)

Documents

40057

Supelco

2,4-Dinitrophenol solution

reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

reference material

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H4N2O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H

InChI key

UFBJCMHMOXMLKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L V Grubskaia et al.
Ukrains'kyi biokhimichnyi zhurnal (1999 ), 84(1), 45-52 (2012-06-12)
The influence of ionizing radiation with low absorbed dose rate (55 mGy x min(-1)) in 1, 12 and 24 hours after irradiation in doses of 0.1; 0.5 and 1.0 Gy on functional state of the electron transfer chain of the
K Srinivasan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 94, 89-100 (2012-04-21)
The formation of host-guest inclusion complex of 2,4-dinitrophenol (2,4-DNP) with nano-hydrophobic cavity of β-cyclodextrin (β-CD) in solution phase was studied by UV-visible spectrophotometer and electrochemical method (cyclic voltammetry, CV). The prototropic behaviors of 2,4-DNP with and without β-CD and the
Asish Mandal et al.
Applied biochemistry and biotechnology, 167(5), 1052-1060 (2012-01-10)
Microbial xylanases have a promising biotechnological potential to be used in industries. In this study, regulation of xylanase production was examined in Bacillus cereus BSA1. Xylanase production was induced by xylan. The enzyme production further increased in the presence of
Francesco Marotta et al.
Rejuvenation research, 15(2), 153-156 (2012-04-27)
The aim of this study was to evaluate the effect of DTS-phytocompound on oxidant-antioxidant balance and protein damage in the kidneys of rats administered high doses of fructose. Adult male Wistar rats were divided into four groups. Group A received
Peripheral neuropathy due to dinitrophenol used for weight loss: something old, something new.
Lauren Phillips et al.
Neurology, 80(8), 773-774 (2013-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service