Skip to Content
Merck
All Photos(1)

Key Documents

28399-U

Supelco

Dual-Layer Carbon Reversible Tube (Micro-Column)

for use with Dioxin Sample Preparation Kit, O.D. 6.35-10 mm

Synonym(s):

Carbon Reversible SPE cartridge tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NB.21

material

glass hardware
polyethylene frit
wire screens (hold carbon layers and frits in place)

Quality Level

Agency

suitable for EPA 1613

composition

bed wt., 100 mg (per carbon layer)

packaging

pk of 10

technique(s)

solid phase extraction (SPE): suitable

O.D.

6.35-10 mm

surface area

1200 m2/g , for Carboxen 1000
75 m2/g , for Carboxen 1016

application(s)

food and beverages

compatibility

for use with 6.35/10mm Reducing Union (Cat. No. 28398-U)
for use with Dioxin Sample Preparation Kit

General description

Dual-Layer Carbon Reversible Tube (Micro-Column) consists of two carbon layers with different affinities for the target compounds and is suitable for use with solid phase extraction (SPE) techniques.

It is used in the Dioxin preparation system for the isolation and separation of polychlorinated biphenyls (PCBs), dioxins, and furan groups based on the different affinities of these compounds for the two carbon layers.

Application

Dual-LayerCarbon Reversible Tube (Micro-Column) has been used in the purification of polychlorinated dibenzodioxins and polychlorinated dibenzofurans (PCDD/Fs) extracted from PUR foam by ultrasound-assisted extraction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of the lignocellulosic filler on the reduction of fire hazard of styrene-butadiene rubber composites, including the reduction of smoke, PCDD/F, PAH emissions and toxicity during its thermal decomposition.
Przemysaw R, et al.
Polymer Testing, 118, 107900-107900 (2023)
Nuevas columnas SLB-5ms de bajo sangrado de Supelco para metodos de compuestos semivola tiles de la EPA de EE. UU.
Katherine K and Stenerson
The Reporter, 17-18 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service