27226
Stoppers
septum diam. 13 mm, red rubber, pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
red rubber
packaging
pkg of 100 ea
O.D.
13 mm
septum diam.
13 mm
compatibility
for use with 7 mL screw top vials
Looking for similar products? Visit Product Comparison Guide
General description
Red rubber stoppers are generally used to seal samples in vials.
Application
It may be used in a comparative study performed to understand the gas tightness between crimp cap vial with a red natural rubber stopper, grey butyl rubber stopper and a screw-cap vial with a grey chlorobutyl rubber stopper.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service