25747
Intermediate Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.25 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
fused silica tubing
Quality Level
composition
phenyl/methyl (deactivated)
packaging
pkg of 1 ea
parameter
360 °C max. temp.
technique(s)
gas chromatography (GC): suitable
L × I.D.
5.0 m × 0.25 mm
Looking for similar products? Visit Product Comparison Guide
General description
Deactivated intermediate polar fused silica tubing can be used as transfer lines, guard columns, or retention gaps, or to make your own columns. It is ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene.
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service