Skip to Content
Merck
All Photos(1)

Key Documents

21516

Swagelok® Reducer

Swagelok® 400-R-2, brass, 1/4 in. Swagelok, for 1/8 in. tube

Synonym(s):

Swagelok® 400-R-2,BRASS REDUCER 1/4X1/8 PK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115717

material

brass

packaging

pkg of 2 ea

manufacturer/tradename

Swagelok® 400-R-2

fitting

for 1/8 in. tube
1/4 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Nuts and ferrules included.

Application

Swagelok® reducer is suitable to be placed on the ends of the tube for sampling ports used for decompositional odor analysis in the following studies:
  • Sampling tubes for graves using gas chromatography-mass spectrometry (GC-MS).
  • Sampling for Burial sites using Triple sorbent traps.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arpad A Vass et al.
Journal of forensic sciences, 53(2), 384-391 (2008-03-28)
This study, conducted at the University of Tennessee's Anthropological Research Facility (ARF), lists and ranks the primary chemical constituents which define the odor of decomposition of human remains as detected at the soil surface of shallow burial sites. Triple sorbent
Arpad A Vass et al.
Journal of forensic sciences, 49(4), 760-769 (2004-08-20)
This study, conducted at the University of Tennessee's Anthropological Research Facility (ARF), describes the establishment of the Decompositional Odor Analysis (DOA) Database for the purpose of developing a man-portable, chemical sensor capable of detecting clandestine burial sites of human remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service