20434
SNOOP® Liquid Leak Detector
8 oz. (236 mL) bottle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
gas chromatography (GC): suitable
Looking for similar products? Visit Product Comparison Guide
General description
Sustained bubble action works even on very small leaks and vertical surfaces. Flexible snooper tube extends for hard-to-reach areas. Does not contain chlorine, aliphatic amines, or ammonium compounds. Dries clean, without staining.
Other Notes
For use with volumetric (bubble) flowmeters. Also useful for checking gas delivery system plumbing for leaks. Not recommend for use upstream of capillary GC systems (an electronic leak detector is preferred).
Legal Information
SNOOP is a registered trademark of Swagelok Company
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service