Skip to Content
Merck
All Photos(1)

Documents

20308

Supelco

Purge/Trap I Vocarb® 4000

for use with Tekmar® Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in./30.5 cm, Swagelok fitting), HT-3

Synonym(s):

Vocarb® 4000 Purge Trap

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

Agency

suitable for EPA 601

Quality Level

manufacturer/tradename

Supelco Vocarb® 4000

application(s)

environmental

compatibility

for use with Tekmar® Velocity XPT, LSC-1, LSC-2, LSC-2000, 4000 (Straight, 12 in./30.5 cm, Swagelok fitting), HT-3

Looking for similar products? Visit Product Comparison Guide

General description

Purge-and-trap is the method of choice for extracting and concentrating volatile organic compounds (VOCs) from almost any matrix. US EPA specifies guidelines for using purge-and-trap methods to process soil, sediment, and water samples. The technique is also used to extract VOCs from foods, beverages, vegetation, pharmaceuticals, petrochemicals, etc. The tubes used in purge-and-trap analyses are generally packed with multiple beds of various sorbent materials, so that a broad range of high and low molecular weight compounds, polar and nonpolar, can be trapped in a single tube.

Legal Information

Tekmar is a registered trademark of Tekmar Co.
Vocarb is a registered trademark of Siemens Industry, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service