Skip to Content
Merck
All Photos(13)

Key Documents

ZRB2165

Sigma-Aldrich

Anti-PRRX1 Antibody, clone 1L16 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Paired mesoderm homeobox protein 1;Homeobox protein PHOX1;Paired-related homeobox protein 1;PRX-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L16, recombinant monoclonal

description

1L16 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 24.3 kDa
observed mol wt ~30-33 kDa

purified by

using Protein A

species reactivity

rat, human

species reactivity (predicted by homology)

mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PRRX1(5396)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1L16 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Paired mesoderm homeobox protein 1 (PRRX1). It targets an epitope within 21 amino acids from the N-terminal half. Predicted to react with both isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal half of human Paired mesoderm homeobox protein 1 (PRRX1).

Application

Quality Control Testing

Evaluated by Western Blotting in C6 cell lysate.

Western Blotting Analysis: 1:10,000 of this antibody detected PRRX1 in C6 cell lysate.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PRRX1 in C6 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PRRX1 in human testis tissue sections.

Western Blotting Analysis: A representative lot detected PRRX1 in Western Blotting applications (Data Courtesy of Dr Takarada from Okayama University).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Paired mesoderm homeobox protein 1 (UniProt: P54821; also known as Homeobox protein PHOX1, Paired-related homeobox protein 1, PRX-1) is encoded by the PRRX1 (also known as PMX1) gene (Gene ID: 5396) in human. PRRX1 is considered as a master transcription factor in mesenchymal cells in embryos and in certain cancer types. It can serve as a transcription co-activator, enhancing the DNA-binding activity of serum response factor that is required for the induction of genes by growth and differentiation factors. PRRX1 acts as a transcriptional regulator of muscle creatine kinase and has a role in the establishment of diverse mesodermal muscle types. It binds to an A/T-rich element in the muscle creatine enhancer. PRRX1 is reported to orchestrates the functional drift of fibroblasts into myofibroblastic phenotype via TGF-β signaling. These reprogrammed fibroblasts have myofibroblastic functions that result in enhanced tumorigenicity and aggressiveness of cancer. PRRX1 expression in cancer-associated fibroblast (CAF) is reported to have an unfavorable prognosis in multiple cancer types. Fibroblast-specific PRRX1 depletion can induce long-term and sustained complete remission of chemotherapy-resistant cancer in murine models. PRRX1 expression is also observed in quiescent human oligodendrocyte progenitor cells (OPCs) where it induces cell-cycle arrest and inhibits migration. Enforced PRRX1 expression can result in reversible G1/0 arrest. Two isoforms of PRRX1 have been described that are produced by alternative splicing. Although both isoforms are known to reduce OPCs proliferation, only PRRX1a reduces migration. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Lee, KW., et al. (2022). Nat. Commun. 13; Article 2793; Wang, J., et al. (2018). Cell Reports 25; 3435-3450).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service