Skip to Content
Merck
All Photos(11)

Key Documents

ZRB1144

Sigma-Aldrich

Anti-IKB α Antibody, clone 6O8, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6O8, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 35.61 kDa
observed mol wt ~37 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NFKBIA(4792)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 6O8 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects I-kappa-B-alpha. It targets an epitope with in the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 143 amino acids from the N-terminal half of human I-kappa-B-alpha.

Application

Anti-IKB alpha, clone 6O8 ZooMAb , Cat. No. ZRB1144, is a recombinant rabbit monoclonal antibody that specifically targets IKB alpha and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected IKB alpha in human stomach tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected IKB alpha in human stomach tissue sections.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

NF-kappa-B inhibitor alpha (UniProt: P25963; also known as I-kappa-B-alpha, IkB-alpha, IkappaB alpha, Major histocompatibility complex enhancer-binding protein MAD3) is encoded by the NFKBIA (also known as IKBA, MAD3, NFKBI) gene (Gene ID: 4792) in human. IkB is known to inhibit the activity of dimeric NF-kappa-B/REL complexes by trapping Rel dimers in the cytoplasm through masking of their nuclear localization signals. In most resting cells, NF-kB is sequestered in the cytoplasm in an inactive form associated with IkB, which blocks the ability of NF-kB to translocate to the nucleus. However, following exposure to pro-inflammatory cytokines, UV light, reactive oxygen species, or bacterial and viral toxins, IkB is phosphorylated by IkB kinases (IKK) and is degraded in the proteasomes. This unmasks the nuclear localization signal of NF-kB and allows it to translocate to the nucleus where it binds to NF-kB response elements in target genes and regulates their transcription. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrián Jordá et al.
International journal of molecular sciences, 23(19) (2022-10-15)
Ranolazine (Rn) is a drug used to treat persistent chronic coronary ischemia. It has also been shown to have therapeutic benefits on the central nervous system and an anti-diabetic effect by lowering blood glucose levels; however, no effects of Rn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service