ZRB1012
Anti-Choline Acetyltransferase (ChAT) Antibody, clone 2K16, ZooMAb® Rabbit Monoclonal
![Enhanced Validation antibodies are tested to ensure reproducibility, specificity, and performance using our enhanced validation strategies enhanced validation](/static/enhanced_validation_badge.png)
recombinant, expressed in HEK 293 cells
Synonym(s):
CHOACTase, ChAT, Choline acetylase, EC: 2.3.1.6
About This Item
Recommended Products
biological source
rabbit (recombinant)
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2K16, recombinant monoclonal
monoclonal
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 82.54 kDa
observed mol wt ~65 kDa
species reactivity
rat, mouse
species reactivity (predicted by homology)
human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
immunofluorescence: suitable using 1:100
immunohistochemistry: suitable using 1:100
western blot: suitable using 1:1,000
isotype
IgG
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CHAT(1103)
General description
Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.
Learn more about ZooMAb here.
Specificity
Immunogen
Application
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Choline Acetyltransferase (ChAT) in rat brain tissue sections.
Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected Choline Acetyltransferase (ChAT) in mouse brain tissue sections.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service