Key Documents
Z740158
LifeSep magnetic separator unit
96P, for 96 well tray, deep well
Synonym(s):
cell sorting magnetic seperation unit
About This Item
UNSPSC Code:
41100000
NACRES:
NA.56
feature
deep well
for 96 well tray
manufacturer/tradename
Dexter Magnetic Technologies 2501005
Related Categories
General description
Durable, easy-to-use and designed to accommodate industry standard microtitre plates. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
Features and Benefits
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Optimized field gradient within wells
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
Choose from one of the most recent versions:
Certificates of Analysis (COA)