Skip to Content
Merck
All Photos(3)

Key Documents

Z725072

SealMate system for adhesive microplate-sealing films

AeraSeal starter kit, includes SealMate dispenser + 2 rolls of AeraSeal film, sterile

Synonym(s):

microplate sealing film, multiwell plate sealing film, sealing film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22
Pricing and availability is not currently available.

material

clear polypropylene plate
clear wells
polyester film

sterility

sterile

packaging

pack of 1 ea (Dispenser plus 2 rolls of film)

manufacturer/tradename

Excel Scientific SM-KIT-BS

W × L

7.9 cm × 10 m

size

Up to 1536 wells

color

white

Looking for similar products? Visit Product Comparison Guide

General description

Excel Scientific′s unique SealMate system provides a convenient new method for sealing standard-format microplates with adhesive films. The system includes a molded polypropylene dispenser with a hinged "clamshell" design and sealing films supplied in a novel sheeted-roll configuration.

The SealMate dispenser provides a protective enclosure for films on the roll prior to dispensing. It can be opened, permitting insertion of a new roll, then closed and latched securely by means of clasps at each side of its film-dispensing "bill". ndividual films on the roll are delineated by non-adhesive color bands that serve as end-tabs for positioning the films on the microplates. The bands also act as markers to show when the end of one film has been reached so that the next film is not pulled prematurely from the dispenser. Each non-adhesive band contains two perforations, the first of which, bounded by a small indent at the edges, tears easily to separate the sheet being dispensed from the next sheet on the roll. The second perforation permits the end-tab to be removed from the plate once the film has been applied. Several unique features contribute to ease and usability of the SealMate dispenser. A compression-brake mechanism prevents the roll from turning as the dispensed sheet is separated from the roll. The smooth lower surface can be swept across the film once applied to the plate to assure secure sealing around all wells. Thumb and forefinger notches above and below the film at the edge of the dispensing bill provide access to the non-adhesive band to facilitate pulling the next film from the dispenser. A prop on the lower surface prevents the bill from contacting the supporting surface when the dispenser is not in use. Alternatively, the dispenser can stand on one of the hubs at the roll ends during storage. The dispenser, thus parked, provides protection and keeps unused films handy for sealing subsequent plates without the need for opening small boxes or pulling individual films from plastic bags. The SealMate system reduces laboratory waste because the film is rolled on itself with no extra paper or plastic liner to be discarded before sealing the plate.

SealPlate® Films

SealPlate® 38 μm thick polyester films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal, not just a cover. They are recommended for ELISA tests, general incubation, and short-term storage. Temperature range: -40 °C to +120 °C. SealPlate® films are non-pierceable. Each SealPlate® roll contains films to cover 100 microplates.
Color code: green

AeraSeal Films

AeraSeal 140 μm thick hydrophobic porous films with medical-grade adhesive provide non-cytotoxic breathable closures for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cellor bacterial growth. AeraSeal films allow uniform air and CO2 exchange for all wells, unlike plate lids which favor exchange for wells near plate edges. Temperature range: -20 °C to +80 °C. AeraSeal films are easily pierceable with pipets or pipet tips for sample recovery. Each AeraSeal roll contains films to cover 50 culture plates.
Color code: red

Features and Benefits

  • convenient, easy film application for microplates
  • three times faster than sheet films
  • inexpensive applicator
  • reduced waste; no protective backing to remove and discard
  • smooth lower surface for pressing film to the plate
  • available sterile and non-sterile
  • seals color coded for identification: green = SealPlate® films; red = AeraSeal films

Legal Information

AeraSeal is a trademark of Excel Scientific, Inc.
SealMate is a trademark of Excel Scientific, Inc.
SealPlate is a registered trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service