Key Documents
Z681784
Culture tubes
size 25 mm × 150 mm , closure caps are sold separately, borosilicate glass tube
Synonym(s):
culture tube, test tube, test tubes
About This Item
UNSPSC Code:
41121706
NACRES:
NB.22
Pricing and availability is not currently available.
material
borosilicate glass tube
conical cap (Push fit)
round bottom
straight neck
transparent tube
sterility
non-sterile
feature
neck thread push fit
packaging
case of 288 ea (4 x 72)
pack of 72 ea
manufacturer/tradename
Kimble® 45048-25150
capacity
55 mL
size
25 mm × 150 mm , closure caps are sold separately
General description
KIMAX® culture tube has a fire-polished top rim and uniform wall thickness for maximum heat transfer and chemical resistance.
Features and Benefits
  • Kimble® culture tubes are manufactured from low extractable, chemical-resistant borosilicate glass.
  • These reusable tubes feature a fire-polished top rim.
  • Uniform wall thickness ensures maximum heat transfer.
  • The marking spot is printed in durable white ceramic enamel.
Packaging
Packaged in a sturdy cardboard box with internal grid to minimize damage during shipping and handling.
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Questions
  1. Can you use metal caps ( diameter of 15-16mm ) as a cap for this specific type of tube? I am looking for culture tubes with cap that can be reused and autoclaved together (no plastic waste).
    1 answer
    1. These tubes are not threaded. The recommended push caps are polypropylene and autoclavable. Unfortunately, a metal push cap is not currently available. Please see the link below to review the cap options for this item:
      https://www.sigmaaldrich.com/US/en/substance/closuresfor25mmculturetubesdisposable1234598765
      Helpful?