Z666106
Greiner centrifuge tubes
capacity 6.5 mL, polystyrene tube, clear, sterile
Synonym(s):
plastic culture tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
Recommended Products
material
polystyrene tube
sterility
sterile
feature
graduations
packaging
case of 1800
manufacturer/tradename
Greiner 151101
capacity
6.5 mL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Clear, round bottom test tubes for sample transfer. Made from polystyrene these tubes are ideally suited for optical measurements as a result of its high clarity.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service