Z657999
Hettich® centrifuge rotors
for ROTANTA 460/460R Centrifuge, Swing-out Rotor, 6 place, to hold, 6 x 5092 buckets
All Photos(1)
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
Recommended Products
feature
capacity 1.5 ‑ 100 mL
to hold 6 x 5092 buckets
packaging
pack of 1 ea
manufacturer/tradename
Hettich 4446
parameter
3,631 max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models ROTANTA 460/460R
Related Categories
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
required but not provided
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)