Skip to Content
Merck
All Photos(2)

Key Documents

Y4895

Sigma-Aldrich

YM-58483

≥98% (HPLC)

Synonym(s):

4-methyl-4′-[3,5-bis(trifluoromethyl)-1H-pyrazol-1-yl]-1,2,3-thiadiazole-5-carboxanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H9F6N5OS
CAS Number:
Molecular Weight:
421.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cc1nnsc1C(=O)Nc2ccc(cc2)-n3nc(cc3C(F)(F)F)C(F)(F)F

InChI

1S/C15H9F6N5OS/c1-7-12(28-25-23-7)13(27)22-8-2-4-9(5-3-8)26-11(15(19,20)21)6-10(24-26)14(16,17)18/h2-6H,1H3,(H,22,27)

InChI key

XPRZIORDEVHURQ-UHFFFAOYSA-N

Application

YM-58483 has been used in the inhibition studies of calcium channels present in cardiomyocytes and pulmonary arterial endothelial cells.

Biochem/physiol Actions

YM-58483 is a selective Ca++ release-activated Ca++ (CRAC) [a.k.a. store-operated Ca++ (SOC) channels] that mediate Ca++ influx in T cells and other inflammatory cells. CRAC channel inhibitors have potential clinical applications in autoimmune disease, transplant rejection, allergic airway inflammation and bronchial asthma. YM-58483 is also promoted as a tool for CRAC studies; may be used as a gold standard.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sodium entry through endothelial store-operated calcium entry channels: regulation by Orai1
Xu N, et al.
American Journal of Physiology. Cell Physiology, 308(4), C277-C288 (2014)
Data on calcium increases depending on stretch in dystrophic cardiomyocytes
Aguettaz E et al.
Data in Brief, 8, 1443-1447 (2016)
Sergei G Gaidin et al.
Glia, 68(6), 1114-1130 (2019-12-20)
Norepinephrine is one of the key neurotransmitters in the hippocampus, but its role in the functioning of the neuroglial networks remains unclear. Here we show that norepinephrine suppresses NH4 Cl-induced oscillations of the intracellular Ca2+ concentration ([Ca2+ ]i ) in
Estella Zuccolo et al.
Stem cells and development, 27(1), 23-34 (2017-11-11)
Stromal cell-derived factor-1α (SDF-1α) drives endothelial colony-forming cell (ECFC) homing and incorporation within neovessels, thereby restoring tissue perfusion in ischemic tissues and favoring tumor vascularization and metastasis. SDF-1α stimulates ECFC migration by activating the G
Marilena Letizia et al.
EMBO molecular medicine, 14(9), e15687-e15687 (2022-08-04)
Inflammatory bowel disease (IBD) is characterized by dysregulated intestinal immune responses. Using mass cytometry (CyTOF) to analyze the immune cell composition in the lamina propria (LP) of patients with ulcerative colitis (UC) and Crohn's disease (CD), we observed an enrichment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service