WPB01525WA
Whirl-Pak® Homogenizer Blender Filter Bag
W × L 38 cm × 38 cm , 15" x 15"
All Photos(1)
About This Item
Recommended Products
sterility
sterile
feature
filter 330 μm
packaging
pkg of 100 ea
manufacturer/tradename
Nasco B01525WA
W × L
38 cm × 38 cm , 15" x 15"
application(s)
sample preparation
compatibility
for use with Whirl-pak® homogenizer
General description
These special bags contain a third middle layer of finely perforated polyethylene, which filters out the solids in the sample when used in a homogenizer blender. Bags feature a filter layer of finely perforated polyethylene, to separate the liquid and solids, allowing for easy pipetting of the sample. The filter is sealed into the perimeter of the bag on the sides and bottom, allowing the sample to be placed in the bag on either side. The liquid will transfer through the filter, but the solids stay on one side.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F, 82° C. Bags can be frozen to any temperature, even liquid nitrogen - careful handling required after freezing. All bags are sterilized after manufacturing.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F, 82° C. Bags can be frozen to any temperature, even liquid nitrogen - careful handling required after freezing. All bags are sterilized after manufacturing.
Legal Information
Whirl-pak is a registered trademark of The Aristotle Corporation