Skip to Content
Merck
All Photos(3)

Key Documents

WH0007124M2

Sigma-Aldrich

Monoclonal Anti-TNF antibody produced in mouse

clone 1C3-A1-F4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DIF, Anti-TNFA, Anti-TNFSF2, Anti-TNFalpha, Anti-tumor necrosis factor (TNF superfamily, member 2)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C3-A1-F4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNF(7124)

Related Categories

General description

This gene encodes a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophages. It can bind to, and thus functions through its receptors TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. This cytokine is involved in the regulation of a wide spectrum of biological processes including cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer. Knockout studies in mice also suggested the neuroprotective function of this cytokine. (provided by RefSeq)
Tumor necrosis factor-alpha (TNF-α) is a homotrimeric proinflammatory cytokine. This gene is located on human chromosome 6. It is a glial-cell related factor. TNF-α belongs to the TNF (tumor necrosis factor) family.

Immunogen

TNF (AAH28148, 1 a.a. ~ 233 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MSTESMIRDVELAEEALPKKTGGPQGSRRCLFLSLFSFLIVAGATTLFCLLHFGVIGPQREEFPRDLSLISPLAQAVRSSSRTPSDKPVAHVVANPQAEGQLQWLNRRANALLANGVELRDNQLVVPSEGLYLIYSQVLFKGQGCPSTHVLLTHTISRIAVSYQTKVNLLSAIKSPCQRETPEGAEAKPWYEPIYLGGVFQLEKGDRLSAEINRPDYLDFAESGQVYFGIIAL

Biochem/physiol Actions

Tumor necrosis factor-alpha (TNF-α) is important in the host immune response to infection. It act as a mediator in severe periportal fibrosis (PPF). TNF-α stimulates apoptosis. This cytokine is linked with the pathogenesis of many autoimmune and inflammatory diseases.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association between TNF-?-238G/A gene polymorphism and OCD susceptibility: A meta-analysis
Jiang C, et al.
Medicine (2018)
Tumor necrosis factor-alpha (TNF-alpha) increases both in the brain and in the cerebrospinal fluid from parkinsonian patients
Mogi M, et al.
Neuroscience Letters, 165(1-2), 208-210 (1994)
Influence of a TNF-? Polymorphism on the Severity of Schistosomiasis Periportal Fibrosis in the Northeast of Brazil
Silva PCV, et al.
Genetic Testing and Molecular Biomarkers, 21(11), 658-662 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service