Skip to Content
Merck
All Photos(6)

Documents

WH0005175M1

Sigma-Aldrich

Monoclonal Anti-PECAM1 antibody produced in mouse

clone 1D2-1A5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD31, Anti-PECAM1, Anti-platelet/endothelial cell adhesion molecule (CD31 antigen)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D2-1A5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

General description

The platelet and endothelial cell adhesion molecule 1 (PECAM1) gene, spanning 75-kb, is mapped on the long arm of human chromosome 17. This gene codes for 130kDa protein, which belongs to the immunoglobulin (Ig) superfamily. It is a vital component of the endothelial cell intercellular junction. PECAM1 is expressed on the surface of circulating platelets, monocytes, neutrophils, and selected T-cell subsets.

Immunogen

PECAM1 (AAH22512, 29 a.a. ~ 738 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ENSFTINSVDMKSLPDWTVQNGKNLTLQCFADVSTTSHVKPQHQMLFYKDDVLFYNISSMKSTESYFIPEVRIYDSGTYKCTVIVNNKEKTTAEYQVLVEGVPSPRVTLDKKEAIQGGIVRVNCSVPEEKAPIHFTIEKLELNEKMVKLKREKNSRDQNFVILEFPVEEQDRVLSFRCQARIISGIHMQTSESTKSELVTVTESFSTPKFHISPTGMIMEGAQLHIKCTIQVTHLAQEFPEIIIQKDKAIVAHNRHGNKAVYSVMAMVEHSGNYTCKVESSRISKVSSIVVNITELFSKPELESSFTHLDQGERLNLSCSIPGAPPANFTIQKEDTIVSQTQDFTKIASKSDSGTYICTAGIDKVVKKSNTVQIVVCEMLSQPRISYDAQFEVIKGQTIEVRCESISGTLPISYQLLKTSKVLENSTKNSNDPAVFKDNPTEDVEYQCVADNCHSHAKMLSEVLRVKVIAPVDEVQISILSSKVVESGEDIVLQCAVNEGSGPITYKFYREKEGKPFYQMTSNATQAFWTKQKANKEQEGEYYCTAFNRANHASSVPRSKILTVRVILAPWKKGLIAVVIIGVIIALLIIAAKCYFLRKAKAKQMPVEMSRPAVPLLNSNNEKMSDPNMEANSHYGHNDDVGNHAMKPINDNKEPLNSDVQYTEVQVSSAESHKDLGKKDTETVYSEVRKAVPDAVESRYSRTEGSLDGT

Biochem/physiol Actions

Platelet and endothelial cell adhesion molecule 1 (PECAM1) also called as cluster of differentiation 31 (CD31), plays a crucial role in the adhesion cascade leading to leukocyte extravasation during the inflammatory process. PECAM1 is essential for leukocyte transmigration via intercellular junctions of vascular endothelial cells. Genetic polymorphism of PECAM1 is associated with atherosclerotic events. PECAM1, is a membrane glycoprotein that facilitates both homophilic and heterophilic adhesion. PECAM1 is also implicated in angiogenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiankun Wang et al.
Experimental and therapeutic medicine, 21(2), 160-160 (2021-01-19)
Mineral smectite granules have traditionally been used for diarrhea and gastrointestinal bleeding; however, the wound healing ability of mineral smectite granules has not been investigated in detail. Therefore, the present study explored the efficacy and safety of mineral smectite granules
Huanhuan Joyce Chen et al.
Methods in molecular biology (Clifton, N.J.), 1907, 91-102 (2018-12-14)
The tissue engineering method of decellularization and recellularization has been successfully used in a variety of regenerative medicine applications. The protocols used to de/recellularize various organs and tissues are largely different. Here we describe a method to effectively engineer a
H M DeLisser et al.
The American journal of pathology, 151(3), 671-677 (1997-09-01)
The adhesive interactions of endothelial cells with each other and the adhesion receptors that mediate these interactions are probably of fundamental importance to the process of angiogenesis. We therefore studied the effect of inhibiting the function of the endothelial cell-cell
The biology of PECAM-1.
P J Newman
The Journal of clinical investigation, 99(1), 3-8 (1997-01-01)
W A Muller et al.
The Journal of experimental medicine, 178(2), 449-460 (1993-08-01)
Platelet/endothelial cell adhesion molecule 1 (PECAM-1; CD31) is crucial to the process of leukocyte transmigration through intercellular junctions of vascular endothelial cells. A monoclonal antibody to PECAM, or recombinant soluble PECAM, blocks transendothelial migration of monocytes by 70-90%. Pretreating either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service