Skip to Content
Merck
All Photos(5)

Key Documents

WH0004082M6

Sigma-Aldrich

Monoclonal Anti-MARCKS antibody produced in mouse

clone 2C2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-80KL, Anti-FLJ14368, Anti-MACS, Anti-MRACKS, Anti-PKCSL, Anti-PRKCSL, Anti-myristoylated alanine-rich protein kinase C substrate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C2, monoclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MARCKS(4082)

General description

Myristoylated alanine-rich C-kinase substrate (MARCKS) contains an N-terminal domain (ND), effector domain (ED), and myristoylated MH2 domain. The MARCKS gene is mapped to human chromosome 6q21.

Immunogen

MARCKS (NP_002347, 2 a.a. ~ 65 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GAQFSKTAAKGEAAAERPGEAAVASSPSKANGQENGHVKVNGDASPAAAESGAKEELQANGSAP

Application

Monoclonal Anti-MARCKS antibody produced in mouse has been used in immunohistochemistry (1:400).

Biochem/physiol Actions

Myristoylated alanine-rich C-kinase substrate (MARCKS) is involved in mediating brain plasticity, inflammatory response, embryonic development, and regeneration processes. It also plays a key role in cell cycle regulation and transmembrane transport. Various studies have implicated the association of MARCKS with the pathophysiology of glioblastoma, cholangiocarcinoma, melanoma, and many more tumor types.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John J Park et al.
Oncogenesis, 13(1), 9-9 (2024-02-29)
Protein kinase C (PKC) is activated downstream of gain-of-function GNAQ or GNA11 (GNAQ/GNA11) mutations in over 90% of uveal melanoma (UM). Phase I clinical trials of PKC inhibitors have shown modest response rates with no survival benefit in metastatic UM.
Hervé Ghesquieres et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 33(33), 3930-3937 (2015-10-16)
We performed a multistage genome-wide association study to identify inherited genetic variants that predict outcome in diffuse large B-cell lymphoma patients treated with immunochemotherapy. We conducted a meta-analysis of two genome-wide association study data sets, one from the LNH2003B trial
Mohamed El Amri et al.
Journal of biomedical science, 25(1), 43-43 (2018-05-24)
The Myristoylated Alanine-Rich C-kinase Substrate (MARCKS) and MARCKS-like protein 1 (MARCKSL1) have a wide range of functions, ranging from roles in embryonic development to adult brain plasticity and the inflammatory response. Recently, both proteins have also been identified as important
Krista Rombouts et al.
Cancer letters, 333(2), 244-252 (2013-02-05)
Several actin-binding proteins have been shown to be altered in metastatic cell lines and tumours and, in particular, Myristoylated Alanine-Rich protein Kinase C substrate (MARCKS) has been implicated in the pathogenesis of various highly metastatic epithelial malignancies. Considering that a
Takeyuki Sugawara et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(26), E5256-E5265 (2017-06-14)
Dendritic spines of Purkinje cells form excitatory synapses with parallel fiber terminals, which are the primary sites for cerebellar synaptic plasticity. Nevertheless, how density and morphology of these spines are properly maintained in mature Purkinje cells is not well understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service