Skip to Content
Merck
All Photos(1)

Key Documents

V6879

Sigma-Aldrich

[Lys8]-Vasopressin

≥98% (HPLC)

Synonym(s):

Lysine vasopressin, Lyspressin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H65N13O12S2
CAS Number:
Molecular Weight:
1056.22
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥98% (HPLC)

form

powder

solubility

0.1 M acetic acid: 20 mg/mL, clear, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(O)=O.NCCCCC(NC(=O)C1CCCN1C(=O)C2CSSCC(N)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(Cc4ccccc4)C(=O)NC(CCC(N)=O)C(=O)NC(CC(N)=O)C(=O)N2)C(=O)NCC(N)=O

InChI

1S/C46H65N13O12S2.C2H4O2/c47-17-5-4-9-29(40(65)52-22-38(51)63)54-45(70)35-10-6-18-59(35)46(71)34-24-73-72-23-28(48)39(64)55-31(20-26-11-13-27(60)14-12-26)43(68)56-32(19-25-7-2-1-3-8-25)42(67)53-30(15-16-36(49)61)41(66)57-33(21-37(50)62)44(69)58-34;1-2(3)4/h1-3,7-8,11-14,28-35,60H,4-6,9-10,15-24,47-48H2,(H2,49,61)(H2,50,62)(H2,51,63)(H,52,65)(H,53,67)(H,54,70)(H,55,64)(H,56,68)(H,57,66)(H,58,69);1H3,(H,3,4)

InChI key

NKWAIKZITYLOMT-UHFFFAOYSA-N

Gene Information

human ... AVP(551)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Lys-Gly-NH2 [Disulfide Bridge: 1-6]

Application

[Lys8]-Vasopressin was used as a test compound in quantitative analysis of peptides and proteins by using on-line capillary isoelectric focusing-mass spectrometry.The product was used for studying the coupling of a polymer-based microchip electrophoresis to MALDI-TOF MS by using rotating ball interface.

Biochem/physiol Actions

Predominant form of vasopressin present in pigs and marsupials. It has lower vasopressor and antidiuretic activity than Arg8-vasopressin.
[Lys8]-Vasopressin also refers as Lysine vasopressin or Lyspressin is the predominant form of vasopressin present in pigs and marsupials. It antagonizes the amnesia in rats. Lyspressin can also induce resistance to extinction of active avoidance behaviour.

Preparation Note

2 mg of [Lys8]-Vasopressin dissolves in 0.1 mL, 0.1M HOAc to yield a clear, colorless solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vladimír Hrabovský et al.
Wiener klinische Wochenschrift, 124(23-24), 815-821 (2012-11-28)
Hypocholesterolemia is commonly found in critically ill patients; however, the aetiology of this condition remains unclear. Several clinical studies refer to the possible negative impact of haemodilution on cholesterol (CH) metabolism in acute medical conditions. The aim of this study
Chris Corbett et al.
European journal of gastroenterology & hepatology, 25(3), 344-351 (2013-01-29)
Variceal bleeding has a 6-week mortality of 20%. Recent evidence suggests that early covered transjugular intrahepatic portosystemic stent shunts (TIPSS) can improve outcomes following a variceal bleed in selected patients. We aim to assess the outcomes following the insertion of
Yukihiro Kuroda et al.
Journal of pharmaceutical and biomedical analysis, 37(3), 423-428 (2005-03-03)
On-line capillary isoelectric focusing-mass spectrometry (cIEF-MS) was applied to determine concentrations of peptides and proteins using angiotensin II and human tetrasialo-transferrin as the model samples. The concentration of the carrier ampholyte was optimized for both resolution and ion intensity. cIEF-MS
Elsa Solà et al.
Current opinion in organ transplantation, 18(3), 265-270 (2013-05-09)
Hepatorenal syndrome (HRS) is a dreaded complication of end-stage liver disease. The best treatment option for HRS is liver transplantation (LT) in suitable candidates. Pretransplant care of HRS is of utmost importance in order to secure a good posttransplant outcome.
Y-Y Huang et al.
The Journal of international medical research, 40(5), 1735-1744 (2012-12-05)
To investigate the presence of aquaporin (AQP) 1 and AQP2 in kidneys of cirrhotic rats with ascites, and to determine the effect of terlipressin on AQP1 and AQP2 levels and its therapeutic efficacy in ascites treatment. Eighteen rats were randomly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service