Skip to Content
Merck
All Photos(7)

Key Documents

V2876

Sigma-Aldrich

Vitamin B12

≥98%

Synonym(s):

α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
Beilstein:
4122889
EC Number:
UNSPSC Code:
12352205
eCl@ss:
34058004
PubChem Substance ID:
NACRES:
NA.79

Quality Level

Assay

≥98%

form

powder

technique(s)

HPLC: suitable

color

red to brown

storage temp.

2-8°C

SMILES string

C/C(C1=N[C@]([C@]([C@H](CC(N)=O)[C@@]/2(C)CCC(NC[C@@H](OP([O-])(O[C@H]3[C@@H](O)[C@@H](N4C=[N]5C6=C4C=C(C)C(C)=C6)O[C@@H]3CO)=O)C)=O)([H])N([Co+]5C#N)C2=C(C)/C([C@@H](CCC(N)=O)C/7(C)C)=NC7=C/8)(C)[C@@](C)(CC(N)=O)[C@@H]1CCC(N)=O)=C9N=C8[C@@H](CCC(N)=O)[C@

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57?,59-,60+,61+,62+;;/m1../s1

InChI key

RMRCNWBMXRMIRW-QJRSUKKJSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Vitamin B12 has been used:
  • as a constituent in vitamin mix solution in in vitro gut simulator
  • as a standard in size exclusion inductively coupled plasma mass spectroscopy (SEC-ICP-MS)
  • as a constituent of simulated plasma protein mixture for Raman spectra analysis
  • in surface plasmon resonance (SPR) analysis
  • as a medium supplement in neonatal rat ventricular myocytes (NRVM)

Biochem/physiol Actions

Vitamin B12 (cobalamin) refers to a group of chemically-related cobalt containing molecules involved in cell processes such as DNA synthesis, fatty acid synthesis, energy production and regulation. The physiologically active forms of vitamin B12 include methylcobalamin and adenosylcobalamin, whereas hydroxocobalamin (vitamin B12a, OHCbl) and cyanocobalamin (CNCbl) are storage and delivery forms. Bacteria-derived hydroxocobalamin (OHCbl) and CNCbl are converted in humans to useful coenzyme forms to support metabolic processes such as mitochondrial methylmalonylcoenzyme A mutase conversion of methylmalonic acid (MMA) to succinate to link lipid and carbohydrate metabolism, and activation of methionine synthase, the rate limiting step in the synthesis of methionine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recombinant human collagen-based microspheres mitigate cardiac conduction slowing induced by adipose tissue-derived stromal cells
Smit NW, et al.
PLoS ONE, 12(8), e0183481-e0183481 (2017)
Raman spectroscopic analysis of high molecular weight proteins in solution-considerations for sample analysis and data pre-processing
Parachalil DR, et al.
Analyst, 143(24), 5987-5998 (2018)
A missense mutation in TCN2 is associated with decreased risk for congenital heart defects and may increase cellular uptake of vitamin B12 via Megalin
Li P, et al.
Oncotarget, 8(33), 55216-55216 (2017)
Microbially Produced Imidazole Propionate Impairs Insulin Signaling through mTORC1
Koh A, et al.
Cell, 175(4), 947-961 (2018)
Speciation of zinc in fish feed by size exclusion chromatography coupled to inductively coupled plasma mass spectrometry-Using fractional factorial design for method optimization and mild extraction conditions
Silva MS, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1104, 262-268 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service