Skip to Content
Merck
All Photos(1)

Documents

V2501

Sigma-Aldrich

Snake venom from Vipera russelli (Russell′s Viper)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

storage temp.

−20°C

Quality Level

Application

Snake venom from Vipera russelli (Russell′s Viper) has also been used as a positive control in indirect and sandwich enzyme-linked immunosorbent assay (ELISA) to study the performance of the immunochromatographic test (ICT)-Viper in venom detection in vitro and the detection of clinical envenoming, respectively.
Snake venom from Vipera russelli (Russell′s Viper) has been used for extraction of coagulant protein for complex generation with bovine X factor.

Biochem/physiol Actions

Snake venom can impose death on humans and animals. Nevertheless, snake venom also exhibits anti-bacterial and wound healing properties. Therefore, it is used as a therapeutic for treating various diseases including thrombosis, arthritis, and cancer.
Snake venom from Russell′s Viper is rich in toxins and proteinase inhibitors. The receptor from Vipera russelli β-RTX, interacts with monoamines and opiate and prevents their interaction with native receptors. The proteases from Russell′s Viper mediate coagulation in human plasma. The neurotoxicity of the venom is contributed by phospholipases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of lanthanide ions with bovine factor X and their use in the affinity chromatography of the venom coagulant protein of Vipera russelli.
Furie BC and Furie B
The Journal of Biological Chemistry, 250(2), 601-608 (1975)
Snake venom proteins: development into antimicrobial and wound healing agents
PerumalSamy R, et al.
Mini-Reviews in Organic Chemistry, 11(1), 4-14 (2014)
Coagulation factor X activating enzyme from Russell's viper venom (RVV-X). A novel metalloproteinase with disintegrin (platelet aggregation inhibitor)-like and C-type lectin-like domains.
Takeya H, et al.
The Journal of Biological Chemistry, 267(20), 14109-14117 (1992)
Snake Venom Proteinase Inhibitors: II. Chemical Structure of Inhibitor II Isolated from the Venom of Russell's viper (Vipera russelli)
TAKAHASHI H, et al.
Journal of Biochemistry, 76(4), 721-733 (1974)
Characterization and molecular cloning of neurotoxic phospholipases A2 from Taiwan viper (Vipera russelli formosensis)
WANG YM, et al.
European Journal of Biochemistry, 209(2), 635-641 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service