Skip to Content
Merck
All Photos(1)

Key Documents

V1381

Sigma-Aldrich

Methyl trans-vaccenate

≥99% (capillary GC), liquid

Synonym(s):

trans-11-Octadecenoic acid methyl ester, Methyl trans-11-octadecenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O2
CAS Number:
Molecular Weight:
296.49
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99% (capillary GC)

form

liquid

functional group

ester

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]\C(CCCCCC)=C(\[H])CCCCCCCCCC(=O)OC

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h8-9H,3-7,10-18H2,1-2H3/b9-8+

InChI key

PVVODBCDJBGMJL-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dragan Vasilev et al.
Meat science, 162, 108020-108020 (2019-12-08)
The fatty acid (FA) profiles of lamb muscle and adipose tissue originating from ten different grazing areas in North Macedonia and an assessment of whether the meat origin could be distinguished on the basis of FA profile are presented. Muscle
Veronika M Lieb et al.
Journal of agricultural and food chemistry, 65(18), 3617-3626 (2017-04-25)
Morphological traits, total lipid contents, and fatty acid profiles were assessed in fruits of several accessions of Elaeis oleifera [Kunth] Cortés, Elaeis guineensis Jacq., and their interspecific hybrids. The latter featured the highest mesocarp-to-fruit ratios (77.9-78.2%). The total lipid contents
T Wang et al.
Asian-Australasian journal of animal sciences, 26(2), 189-194 (2013-02-01)
The goal of this study was to evaluate the effects of lactation stage and individual performance on milk cis-9, trans-11 conjugated linoleic acid (CLA) content in dairy cows. In experiment 1, the milk cis-9, trans-11 CLA content from dairy cows
D D Bikle et al.
Endocrinology, 114(1), 260-267 (1984-01-01)
To evaluate whether membrane fluidity plays a role in regulating calcium flux across the intestinal brush border, we purified brush border membranes from different regions of the chick intestine and determined the relationship of their ability to transport calcium and
Grazia Pallara et al.
BMC veterinary research, 10, 271-271 (2014-11-27)
Stoned olive pomace (SOP), which represents approximately 50% of the conversion process of olives to olive oil, is largely not utilised and creates costs for its disposal and has negative environmental impacts. In vitro trial experiments were employed to study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service