U4125
Uridine 5′-(trihydrogen diphosphate) sodium salt from Saccharomyces cerevisiae
95-100%
Synonym(s):
UDP sodium salt, Uridine 5′-(trihydrogen pyrophosphate) sodium salt
About This Item
Recommended Products
Assay
95-100%
solubility
H2O: soluble 100 mg/mL, clear, colorless to very faintly yellow
storage temp.
−20°C
SMILES string
[Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])([O-])=O)N2C=CC(=O)NC2=O
InChI
1S/C9H14N2O12P2.3Na/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(22-8)3-21-25(19,20)23-24(16,17)18;;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,10,12,15)(H2,16,17,18);;;/q;3*+1/p-3
InChI key
GFASEGWJDXDCKZ-UHFFFAOYSA-K
Gene Information
human ... LPAR4(2846) , LTB4R(1241) , P2RY1(5028) , P2RY10(27334) , P2RY11(5032) , P2RY12(64805) , P2RY13(53829) , P2RY14(9934) , P2RY2(5029) , P2RY4(5030) , P2RY5(10161) , P2RY6(5031) , P2RY8(286530)
Related Categories
Application
Biochem/physiol Actions
Features and Benefits
Preparation Note
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT SE 2
Target Organs
Eyes,Central nervous system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service