TTRT96P250B-EP
Eppendorf twin.tec® Real-Time PCR Plates 96
Skirted, PCR clean, volume 250 μL, blue plate (white wells), pack of 25
All Photos(1)
About This Item
Recommended Products
material
blue plate (white wells)
polypropylene wells
sterility
non-sterile
feature
skirt
PCR clean
Skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951022003 NA Only
technique(s)
PCR: suitable
size
96 well
volume
250 μL
working volume
250 μL
General description
The white wells of Eppendorf’s twin.tec real-time PCR plates increase the reflection of fluorescent light in real-time PCR (qPCR) and other fluorescence assays. The increased fluorescence from these specialized qPCR plates improves the sensitivity of your assays, while also improving reproducibility and consistency of your results.
Application
Eppendorf® twin.tec® Real-Time PCR Plates 96 are suitable for real-time polymerase chain reaction (PCR) and melting curve analysis
Features and Benefits
- Polypropylene, white wells ensure better reflection of fluorescent light
- Highly stable to mechanical stress
- Effective sealing by raised well rims
- PCR plates feature skirted frames
- Optimal heat transfers due to reduced wall thickness
- Autoclavable at 121 °C for 20 min
- White wells for better reflection
- High mechanical stability
- Raised rims for effective sealing
- Skirted and semi-skirted frames
- Optimal heat transfer due to reduced wall thickness
- Autoclavable (121 °C, 20 min)
- PCR clean and Eppendorf Forensic DNA Grade s available
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG