Skip to Content
Merck
All Photos(1)

Key Documents

TOX1

Sigma-Aldrich

In Vitro Toxicology Assay Kit, MTT based

Synonym(s):

mitochondrial activity assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.84

usage

1 kit sufficient for 1,000 tests

packaging

pkg of 1 kit

storage condition

dry at room temperature

λmax

570 nm

application(s)

cell analysis
detection

detection method

colorimetric

storage temp.

2-8°C

General description

This kit is designed for determining cell number/ cell viability spectrophotometrically as a function of mitochondrial activity in living cells. The MTT method is simple, accurate and yields reproducible results. The key component is (3- [4,5- dimethylthiazol-2-yl]-2,5-diphenyl tetrazolium bromide) or MTT. Solutions of MTT, dissolved in medium or balanced salt solutions without phenol red, are yellowish in color. Mitochondrial dehydrogenases of viable cells cleave the tetrazolium ring, yielding purple formazan crystals which are insoluble in aqueous solutions. Absorbance of converted dye is measured at a wavelength of 570nm.

Application

In Vitro Toxicology Assay Kit, MTT based has been used to perform an (3-[4,5-dimethylthiazole-2-yl]-2,5-diphenyltetrazolium bromide) MTT assay to measure the cytotoxicity of gold nanoparticles (AuNP). It has also been used to examine cell viability in SH-SY5Y cultures in response to 6-hydroxydopamine (6-OHDA) treatment.

Biochem/physiol Actions

Conversion of MTT to a water-insoluble colored formazan derivative which is then solubilized in acidic isopropanol.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - Muta. 2 - Skin Corr. 1 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human renal organic anion transporter 1-dependent uptake and toxicity of mercuric-thiol conjugates in Madin-Darby canine kidney cells
Aslamkhan AG, et al.
Molecular Pharmacology, 63(3), 590-596 (2003)
Dalia Chávez-García et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(6), 2396-2406 (2020-02-06)
Luminescent lanthanide downconversion nanoparticles (DCNPs) provide a combination of high luminescence intensity, sharp emission peaks with narrow bandwidth and a large Stokes' shift, leading to high-performance biomedical applications mainly for imaging. The purpose of this study is to present a
J Carmichael et al.
Cancer research, 47(4), 936-942 (1987-02-15)
Drug sensitivity assays were performed using a variation of a colorimetric [3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)] assay on V79, CHO-AuxB1, CHRC5, NCI-H460, and NCI-H249 cell lines following optimization of experimental conditions for each cell line. Results from this assay were compared with
Jun Wang et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 17(2), 343-351 (2008-12-11)
Spliceosome-mediated RNA trans-splicing has emerged as an exciting mode of RNA therapy. Here we describe a novel trans-splicing strategy, which targets highly abundant pre-mRNAs, to produce therapeutic proteins in vivo. First, we used a pre-trans-splicing molecule (PTM) that mediated trans-splicing
T Mosmann
Journal of immunological methods, 65(1-2), 55-63 (1983-12-16)
A tetrazolium salt has been used to develop a quantitative colorimetric assay for mammalian cell survival and proliferation. The assay detects living, but not dead cells and the signal generated is dependent on the degree of activation of the cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service