Skip to Content
Merck
All Photos(1)

Key Documents

TMO137103

Nunc® MicroWell 96 well polystyrene plates

Nunclon® Delta, black, well volume (400 μL), case of 160 ea

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates, ELISA plates, fluorescence-based cell assay plates, immunoassay plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.15
material:
black
polystyrene
plate L × W:
128 mm × 86 mm
well volume:
(400 μL)
wells:
(96)
working volume:
250 μL
sterility:
sterile
feature:
lid
pinchbar design
Pricing and availability is not currently available.

material

black
polystyrene

sterility

sterile

feature

lid
pinchbar design

packaging

case of 160 ea

manufacturer/tradename

Nunc 137103

plate L × W

128 mm × 86 mm

well volume

(400 μL)

wells

(96)

working volume

250 μL

suitability

suitable for (fluorescence-based cell assays)

Looking for similar products? Visit Product Comparison Guide

General description

  • White plates provide maximum reflection, minimum auto-fluorescence and autoluminescence
  • Black plates provide minimal background and light scatter in fluorescence assays
  • Non-toxic white or black plate is provided with clear lid
  • Raised well rims of flat bottom wells reduce risk of cross contamination
  • Low crosstalk
  • Nunclon® Δ for cell based assays
  • Working volume range: 50-250 μl/well
  • Suitable for fluorescence-based cell assays

Legal Information

MicroWell is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service