Skip to Content
Merck
All Photos(7)

Key Documents

T8660

Sigma-Aldrich

Anti-β-Tubulin III Antibody

enhanced validation

mouse monoclonal, SDL.3D10

Synonym(s):

Anti-CDCBM, Anti-CDCBM1, Anti-CFEOM3, Anti-CFEOM3A, Anti-FEOM3, Anti-TUBB4, Anti-beta-4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

SDL.3D10, monoclonal

contains

15 mM sodium azide

species reactivity

bovine, rat, human, boar

packaging

antibody small pack of 25 μL

enhanced validation

knockout
Learn more about Antibody Enhanced Validation

technique(s)

indirect ELISA: suitable
western blot: 1:400 using a rat brain extract

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB3(10381)
rat ... Tubb3(246118)

General description

Monoclonal Anti-μ Tubulin Isotype III (mouse IgG2b isotype) is derived from the SDL.3D10 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice.
Tubulin β 3 class III (TUBB3) also known as β-Tubulin III, is encoded by the gene mapped to human chromosome 16q24.3. TUBB3 protein expression is restricted to neurons.

Specificity

The antibody specifically recognizes an epitope located on isotype III of β-tubulin. No cross-reactivity with other tubulin isotypes is observed. The antibody can stimulate microtubule assembly when reconstituted with tubulin and either tau or MAP2.

Immunogen

synthetic peptide corresponding to the C-terminal sequence of human β-tubulin isotype III coupled to BSA

Application

It has also been used in immunofluorescence analysis.
Monoclonal Anti-β-Tubulin III antibody produced in mouse has been used in following studies:
  • Immunocytochemistry.
  • Flow cytometry.
  • Immunoblotting.

Biochem/physiol Actions

Tubulin β 3 class III (TUBB3) plays a vital role in nervous system development and axon guidance. Mutation of the gene encoding protein leads to the ocular motility disorder, congenital fibrosis of the extraocular muscle type 3 (CFEOM3), and various neurological syndromes. Altered expression of the protein affects microtubule dynamics and microtubule-kinesin interactions. Increased expression of TUBB3 is observed in tumor tissues. Decreased expression of TUBB3 can be considered as an important marker for poor prognosis of cutaneous malignant melanoma. TUBB3 is implicated in the suppression of invasive growth of tumor tissue;therefore, it is considered to be a potential target for development of antitumor drugs.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Legume lectin FRIL preserves neural progenitor cells in suspension culture in vitro
Yao H, et al.
Clinical & Developmental Immunology, 2008 (2008)
Rebecca M Brown et al.
Journal of virology, 93(17) (2019-06-21)
Congenital human cytomegalovirus (HCMV) infection causes a broad spectrum of central and peripheral nervous system disorders, ranging from microcephaly to hearing loss. These ramifications mandate the study of virus-host interactions in neural cells. Neural progenitor cells are permissive for lytic
Semaphorin CD100 from activated T lymphocytes induces process extension collapse in oligodendrocytes and death of immature neural cells.
Giraudon P
Journal of Immunology, 172(2), 1246-1255 (2004)
Isolation, characterization and multi-lineage differentiation of stem cells from human exfoliated deciduous teeth
Zhang NA, et al.
Molecular Medicine Reports, 14(1), 95-102 (2016)
Anthony J Veloso et al.
ACS chemical neuroscience, 4(6), 924-929 (2013-03-12)
The bioactivities of two novel compounds (TAE-1 and TAE-2) that contain a sym-triazine scaffold with acetylcholine-like substitutions are examined as promising candidate agents against Alzheimer's disease. Inhibition of amyloid-β fibril formation in the presence of Aβ1-42, evaluated by Thioflavin T

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service