Skip to Content
Merck
All Photos(2)

Key Documents

T8276

Sigma-Aldrich

Anti-TRPC1 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Trpc1 Antibody, Trpc1 Antibody - Anti-TRPC1 antibody produced in rabbit, Anti-Transient receptor potential cation channel, subfamily C, member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

rat

technique(s)

immunohistochemistry: suitable
western blot (chemiluminescent): suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRPC1(7220)
mouse ... Trpc1(22063)
rat ... Trpc1(89821)

Related Categories

General description

Transient receptor potential cation channel subfamily C member 1 (TRPC1) is encoded by the gene mapped to human chromosome 3q22-24. The encoded protein is a member of the transient receptor potential (TRP) superfamily of cation channels and is distributed in both glomeruli and glomerular mesangial cells.

Immunogen

Synthetic protein corresponding to amino acids 557-571 of human TRPC1 (GenBank® Accession No. P48995). The epitope is identical in mouse and rat and highly homologous in bovine and Xenopus.

Biochem/physiol Actions

Transient receptor potential cation channel subfamily C member 1 (TRPC1) facilitates Ca2+ entry upon store depletion in a variety of cell types by forming a multimeric complex with other TRPCs. The encoded protein is implicated in the odontoblast-like differentiation of human dental pulp cells (hDPCs). Mutation in the gene is associated with the development type 2 diabetes mellitus (T2D). Decreased expression of the gene leads to hepatocellular carcinoma cell proliferation and thus, TRPC1 can be considered as a potential therapeutic target for hepatocellular carcinoma.

Physical form

Lyophilized at 0.3 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of TRPC1 gene polymorphisms with type 2 diabetes and diabetic nephropathy in Han Chinese population.
Chen K
Endocrine Research, 38, 59-68 (2013)
Wanjie Huang et al.
International journal of molecular medicine, 49(2) (2021-12-23)
Persistent pulmonary hypertension of the newborn (PPHN) is a common pulmonary vascular disease during the neonatal period, and it is associated with a high clinical mortality rate and a poor prognosis. At present, the treatment of PPHN is based mainly
Differential expression of store-operated calcium- and proliferation-related genes in hepatocellular carcinoma cells following TRPC1 ion channel silencing
Selli C
Molecular and Cellular Biochemistry, 420, 129-140 (2016)
The Role of Transient Receptor Potential Cation Channel, Subfamily C, Member 1 in the Odontoblast-like Differentiation of Human Dental Pulp Cells
Song Z
Journal of Endodontics, 43, 315-320 (2017)
Jinwang Ye et al.
Aging cell, 19(9), e13209-e13209 (2020-08-21)
Intracellular accumulating of the hyperphosphorylated tau plays a pivotal role in neurodegeneration of Alzheimer disease (AD), but the mechanisms underlying the gradually aggravated tau hyperphosphorylation remain elusive. Here, we show that increasing intracellular tau could upregulate mRNA and protein levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service