Skip to Content
Merck
All Photos(1)

Key Documents

T6448

Sigma-Aldrich

Anti-TREK-1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Kcnk2, Anti-Potassium Channel K2P2.1, Anti-TWIK-related K+ channel 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

technique(s)

western blot (chemiluminescent): 1:200 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNK2(3776)
rat ... Kcnk2(170899)

General description

Potassium two pore domain channel subfamily K member 2 (KCNK2), also known as TWIK-related K+ channel-1 (TREK1), is expressed in the spinal cord, bladder and dorsal root ganglion. It is part of the TREK family.

Specificity

The antibody reacts with rat TREK-1 (confirmed). It is expected that the antibody will also react with human and mouse TREK-1 due to sequence homology (17/18).

Immunogen

synthetic peptide (DPKSA AQNSK PRLSF STKC) corresponding to the intracellular, amino terminus of human TREK-1 (amino acid residues 8-28).

Application

Anti-TREK-1 antibody produced in rabbit has been used in western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Potassium two pore domain channel subfamily K member 2 (KCNK2) possesses a potassium channel activity. It has a role in stabilizing the resting membrane potential of cells. The protein is also involved in the development of rat heart. TREK-1 could act as a drug target to treat acute heart ischemia.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased Expression of TREK-1 K+ Channel in the Dorsal Root Ganglion of Rats with Detrusor Overactivity After Partial Bladder Outlet Obstruction
Junlong Z,et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 24, 1064-1064 (2018)
Analysis of gene expression profiles in the programmed cell death rat retinal ganglion cells (RGC-5).
Abdelnaby K, et al.
Cancer Therapy, 6(2) (2008)
Effects of fluoxetine on protein expression of potassium ion channels in the brain of chronic mild stress rats
Chunlin C, et al.
Acta Pharmaceutica Sinica. B, 5(1), 55-61 (2015)
Naaz Andharia et al.
Channels (Austin, Tex.), 11(3), 224-235 (2017-01-14)
The TREK-1 channel, the TWIK-1-related potassium (K
Chunlin Chen et al.
Acta pharmaceutica Sinica. B, 5(1), 55-61 (2015-11-19)
The purpose of this study is to investigate the expression of major potassium channel subtypes in the brain of chronical mild stress (CMS) rats and reveal the effects of fluoxetine on the expression of these channels. Rats were exposed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service