Skip to Content
Merck
All Photos(1)

Key Documents

T5200

Sigma-Aldrich

Triparanol

≥97% (HPLC), powder

Synonym(s):

1-(4-(2-Diethylaminoethoxy)phenyl)-1-p-tolyl-2-(4-chlorophenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32NO2Cl
CAS Number:
Molecular Weight:
438.00
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

white to off-white

mp

102.9-103.7 °C

solubility

DMSO: 20 mg/mL
H2O: insoluble
alcohol: soluble
olive oil: slightly soluble

storage temp.

2-8°C

SMILES string

CCN(CC)CCOc1ccc(cc1)C(O)(Cc2ccc(Cl)cc2)c3ccc(C)cc3

InChI

1S/C27H32ClNO2/c1-4-29(5-2)18-19-31-26-16-12-24(13-17-26)27(30,23-10-6-21(3)7-11-23)20-22-8-14-25(28)15-9-22/h6-17,30H,4-5,18-20H2,1-3H3

InChI key

SYHDSBBKRLVLFF-UHFFFAOYSA-N

Gene Information

Application

Triparanol has been used to inhibit cholesterol biosynthesis in lipid rafts1. Triparanol has also been used as a 3β-hydroxysterol-Δ24-reductase inhibitor to study its effect on delayed-rectifier potassium current (Iks) channels2.

Biochem/physiol Actions

Triparanol is known to repress Hedgehog signaling in cancer cells and can also inhibit tumor growth3.
Desmosterol Delta24 (D24) reductase inhibitor.

Preparation Note

Triparanol is soluble in DMSO at 20 mg/ml and is also soluble in alcohol. It is slightly soluble in olive oil and is insoluble in water.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A K Bhattacharyya et al.
Atherosclerosis, 67(1), 33-39 (1987-09-01)
The relationships between the cholesterol content of the diet, plasma cholesterol concentration, cholesterol absorption, and cholesterol synthesis (measured indirectly by desmosterol suppression technique) are explored in groups of high- and low-responding rhesus monkeys fed diets containing 0.02, 0.15 and 0.75
D Raederstorff et al.
Biochimica et biophysica acta, 960(2), 190-199 (1988-05-22)
The tetracyclic sterol precursors, cyclolaudenol, cycloartenol and lanosterol, inhibit efficiently the tetrahymanol biosynthesis in the ciliate Tetrahymena pyriformis, as reported earlier for cholesterol and other sterols. The prokaryotic bacteriohopanetetrols have little effect, and diplopterol, another hopanoid, as well as the
F Echevarria et al.
The Journal of biological chemistry, 265(15), 8484-8489 (1990-05-25)
Zymosterol (5 alpha-cholesta-8(9),24-dien-3 beta-ol) comprised a negligible fraction of the mass of sterol in cultured human fibroblasts but was well labeled biosynthetically with radioactive acetate. Treatment of cells with triparanol, a potent inhibitor of sterol delta 24-reductase, led to a
A Boogaard et al.
The Biochemical journal, 241(2), 345-351 (1987-01-15)
Incubating Hep G2 cells for 18 h with triparanol, buthiobate and low concentrations (less than 0.5 microM) of U18666A, inhibitors of desmosterol delta 24-reductase, of lanosterol 14 alpha-demethylase and of squalene-2,3-epoxide cyclase (EC 5.4.99.7) respectively, resulted in a decrease of
A Ramu et al.
Cancer research, 44(10), 4392-4395 (1984-10-01)
The effects of the triparanol analogues chlorotrianisene, clomiphene, tamoxifen, 5-[p-(fluoren-9-ylidenemethyl)phenyl]-2-piperidineethanol (MDL 10393), MDL 8917v, nafoxidine, 2-[p-(6-methoxy-2-phenylinden-3-yl)phenoxy]triethylamine (U-11555A), 2-[p-(3,4-dihydro-6-methoxy-2-phenyl-1-naphthyl)phenoxy]triethylamine (U-10520A), and nitromifene, as well as triparanol itself, were studied in the P388 murine leukemia cell line and in a doxorubicin-resistant subline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service