Skip to Content
Merck
All Photos(2)

Key Documents

T5125

Sigma-Aldrich

Thimerosal

97.0-101.0% (on dried substance, titration)

Synonym(s):

2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81
Beilstein:
8169555
EC Number:
MDL number:
UNSPSC Code:
51473202
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

97.0-101.0% (on dried substance, titration)

form

powder or crystals

color

white to faint beige

mp

234-237 °C (dec.) (lit.)

solubility

1 g/L at 20 °C

Mode of action

enzyme | inhibits

storage temp.

room temp

SMILES string

[Na+].CC[Hg]Sc1ccccc1C([O-])=O

InChI

1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2

InChI key

RTKIYNMVFMVABJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Thimerosal is a sulfhydryl (thiol) oxidizing reagent capable of inhibiting a wide range of sulfhydryl-dependent enzymes and proteins such as PI3-kinase-dependent methionine synthase, glutamate transport receptors, and Na++K+ dependent ATPases.

Features and Benefits

  • Meets USP/EP testing specifications
  • High-quality chemical, suitable for a wide variety of research applications.

Other Notes

As part of the corporation global plan to support the Minamata Convention eliminating mercury-containing compounds, as of January 2021 customers will be required to provide an End-Use Declaration. The sales of thimerosal will be restricted for approved applications only, such as preservative in vaccine manufacturing, preservative in eye cosmetics, research use within academia (excluding preservation intent) and as reference material only. The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act , European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.
For additional information on our range of Biochemicals, please complete this form.
Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rune Ehrenreich Kuhre et al.
Cell metabolism, 29(3), 719-726 (2018-11-20)
Studies on isolated pancreatic islets suggest that neuromedin U (NMU), a brain and gastrointestinal peptide, acts as a decretin hormone, inhibiting glucose-stimulated insulin secretion. We investigated whether this effect could be reproduced in vivo and in isolated perfused rat pancreas. Unlike
R Chalghoumi et al.
Poultry science, 87(1), 32-40 (2007-12-15)
The present study was an attempt to raise hen egg yolk Ig (IgY) simultaneously directed against Salmonella Enteritidis (SE) and Salmonella Typhimurium (ST) in the same egg yolk. The immunopotentiating effect of 2 different adjuvants -- Freund's adjuvants (FA) and
Alberto Eugenio Tozzi et al.
Developmental medicine and child neurology, 54(9), 843-848 (2012-05-18)
The aim of this article was to explore the effect of duration of breastfeeding on neurocognitive development. The long-term effect of breastfeeding on neurodevelopment was examined through a battery of neuropsychological tests in 1403 children (693 females, 710 males; mean
Liana Basova et al.
iScience, 23(6), 101230-101230 (2020-06-20)
The lacrimal gland (LG) is an exocrine organ responsible for the secretion of aqueous tear film. Regenerative and stem cell therapies that target LG repair are coming to the fore, although our understanding of LG cell lineage hierarchy is still
Sarah K Parker et al.
Pediatrics, 114(3), 793-804 (2004-09-03)
The issue of thimerosal-containing vaccines as a possible cause of autistic spectrum disorders (ASD) and neurodevelopmental disorders (NDDs) has been a controversial topic since 1999. Although most practitioners are familiar with the controversy, many are not familiar with the type

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service