Skip to Content
Merck
All Photos(5)

Key Documents

T3253

Sigma-Aldrich

Trizma® hydrochloride

reagent grade, ≥99.0% (titration), crystalline

Synonym(s):

TRIS HCl, TRIS hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, Tromethane hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3 · HCl
CAS Number:
Molecular Weight:
157.60
Beilstein:
3675235
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

reagent grade

Quality Level

Assay

≥99.0% (titration)

form

crystalline

storage condition

dry at room temperature

technique(s)

affinity chromatography: suitable

impurities

≤0.5% water (Karl Fischer)

color

white

useful pH range

7.0-9.0

pKa (25 °C)

8.1

mp

150-152 °C

solubility

H2O: soluble, clear, colorless

density

1.39 g/cm3 at 20—25 °C
1.41 g/cm3 at 20—25 °C

absorption

≤0.05 at 290 nm at 40%

suitability

suitable for mass spectrometry

application(s)

diagnostic assay manufacturing
life science and biopharma
sample preparation

SMILES string

Cl.NC(CO)(CO)CO

InChI

1S/C4H11NO3.ClH/c5-4(1-6,2-7)3-8;/h6-8H,1-3,5H2;1H

InChI key

QKNYBSVHEMOAJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris HCl, or Trizma® hydrochloride, is a widely utilized acidic buffer in the physiological pH range of 7.3 to 7.5. With a pKa of 8.08 at 25°C, it offers a pH range of 7.0 to 9.0, aligning with living organisms′ typical pH. Essential in various biological, cell biology, and biochemistry applications, Tris HCl is crucial for effective pH control in buffer solutions. Notably, Tris base and Tris hydrochloride alone lack sufficient buffering capacity, prompting their combination within the pH range of 7 to 9.

Tris buffers play a key role in techniques like protein electrophoresis and western blotting, serving in low ionic strength buffers for protein transfer. They are also integral to SDS-PAGE gels, running buffers, and blotting buffers. Additionally, Tris salts, especially in TAE and TBE buffers, are extensively employed in DNA agarose electrophoresis. These buffers are pivotal in diverse applications, including biochemistry, protein purification, and electrophoresis, showcasing the versatility and importance of Tris HCl in maintaining optimal pH for biochemical and cell biology research.

Application

Trizma® hydrochloride has been used as a buffer solution for staining procedure, reduction and alkylation studies and in situ hybridization procedure.­[1][2][3]

Features and Benefits

  • Ideal as a Biological buffer for Cell Biology and Biochemical research
  • Effective Buffering from pH 7-9 with a pKa of 8.1 (25 °C)
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications


Packaging

The DRYPOUR®packaging system for high-quality hygroscopic salts DRYPOUR®packaging:
  • The current DRYPOUR®packaging configuration is in a 50L nestable drum
  • 25KG-DP are the standard prepack offering
  • Smaller volumes can be put in the same drum for variable configurations
The benefits of DRYPOUR®:
  • Improves operational efficiency by reducing material preparation time and increasing operator safety
  • Minimizes caking of hygroscopic salts
  • Offers triple protection: against moisture inside and outside, and against contamination
How does the DRYPOUR®packaging system Improve your operational efficiency?
  • This state-of-the-art design packaging system minimizes caking, thus significantly reducing the time needed for material preparation
  • PE drum with our new tamper-evident seal, a polyethylene liner with integrated desiccant bags and a breathable interior Tyvek®liner
  • Desiccant bags are firmly integrated into the polyethylene liner
  • Triple protection delivers two invaluable results:
  1. Dramatically reduced caking
  2. No contamination risk from the desiccant

Other Notes

For additional information on our range of Biochemicals, please complete this form.
The pH values of all buffers are temperature and concentration dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution. For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

DRYPOUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential effects of delayed aging on phenotype and striatal pathology in a murine model of Huntington disease.
Tallaksen-Greene SJ et al.
The Journal of Neuroscience, 34, 15658-15658 (2014)
Evaluation of the allergenicity and antigenicity of bovine-milk alphas1-casein using extensively purified synthetic peptides.
Elsayed S et al.
Scandinavian Journal of Immunology, 60, 486-486 (2004)
Sandra Lawrynowicz Leibel et al.
Scientific reports, 9(1), 13450-13450 (2019-09-19)
Surfactant protein B (SFTPB) deficiency is a fatal disease affecting newborn infants. Surfactant is produced by alveolar type II cells which can be differentiated in vitro from patient specific induced pluripotent stem cell (iPSC)-derived lung organoids. Here we show the
Tris-tris-HCl: a standard buffer for use in the physiologic pH range.
R A Durst et al.
Clinical chemistry, 18(3), 206-208 (1972-03-01)
Marcel Kuiper et al.
Biotechnology progress, 35(4), e2821-e2821 (2019-04-16)
Perfusion is a cell culture mode that is gaining popularity for the manufacture of monoclonal antibodies and their derivatives. The cell culture media supporting perfusion culture need to support higher cell densities than those used in fed-batch culture. Therefore, when

Questions

1–7 of 7 Questions  
  1. Is this a simple mixture of Tris-base & HCl ? If I want to adjust both the concentration and pH of Tris-HCl, can I achieve the same effect by purchasing two reagents and preparing the solution myself?

    1 answer
    1. This product is trizma® hydrochloride. This product is not a mixture of Trizma® hydrochloride and Trizma® base.

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  4. How can I find the technical bulletin 106B that is referenced in the Product Information Sheet for Product T3253, Trizma® hydrochloride?

    1 answer
    1. Under the Documents section above.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. Is Product T3253, Trizma® hydrochloride, the same as Tris buffer?

    1 answer
    1. No. Trizma HCl in solution at 0.05 M will produce a pH of approximately 4.7, but will have little if any buffering capacity. Trizma Base in solution at 0.05 M will produce a pH of approximately 10.4, but will have little if any buffering capacity. Blending Trizma Base and Trizma HCl will produce any desired pH between 7 and 9, with a reasonable buffering capacity.

      Helpful?

  7. What is the pH of Trizma HCl, Product T3253, in solution?

    1 answer
    1. The pH of Trizma hydrochloride in water is dependent on the concentration. The pH of a 0.1 M aqueous solution is approximately 4.7 while the pH of a 40% (w/w) aqueous solution is typically 2.6-3.3.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service