T3126
D-α-Tocopherol succinate
semisynthetic, 1210 IU/g
Synonym(s):
Vitamin E succinate
About This Item
Recommended Products
biological source
semisynthetic
form
powder
specific activity
1210 IU/g
color
white to yellow
storage temp.
room temp
SMILES string
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(=O)CCC(O)=O)c(C)c(C)c2O1
InChI
1S/C33H54O5/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-20-33(8)21-19-28-27(7)31(25(5)26(6)32(28)38-33)37-30(36)18-17-29(34)35/h22-24H,9-21H2,1-8H3,(H,34,35)/t23-,24-,33-/m1/s1
InChI key
IELOKBJPULMYRW-NJQVLOCASA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- to as a dietary supplement to study its immunomodulatory effects in chicks
- as a dietary supplement in pregnant women to evaluate its effects on arsenic exposure
- as an antioxidant to investigate concurrent pathways of cell death due to acute exercise
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service