Skip to Content
Merck
All Photos(2)

Key Documents

T182

Sigma-Aldrich

Tyrphostin A9

solid

Synonym(s):

Malonoben, [[3,5-bis(1,1-Dimethylethyl)-4-hydroxyphenyl]methylene]propanedinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N2O
CAS Number:
Molecular Weight:
282.38
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (HPLC)

form

solid

color

yellow

mp

139-140 °C

solubility

ethanol: 20 mg/mL
DMSO: <25 mg/mL
H2O: insoluble

SMILES string

CC(C)(C)c1cc(\C=C(\C#N)C#N)cc(c1O)C(C)(C)C

InChI

1S/C18H22N2O/c1-17(2,3)14-8-12(7-13(10-19)11-20)9-15(16(14)21)18(4,5)6/h7-9,21H,1-6H3

InChI key

MZOPWQKISXCCTP-UHFFFAOYSA-N

Application

Tyrphostin A9 can be used for inhibiting tyrosine kinase functions in C2C12 cells. It has also been used to disrupt membrane potential in mammalian cells.

Biochem/physiol Actions

Tyrphostin A9 is a PDGF receptor tyrosine kinase inhibitor that can induce apoptosis in cancer cells, inhibit the growth of vascular smooth cells and block calcium release-dependent phosphorylations.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Preparation Note

Tyrphostin A9 is soluble in ethanol at 20 mg/ml, in DMSO at a concentration less than 25 mg/ml. It is insoluble in water.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyrphostins as molecular tools and potential antiproliferative drugs.
A Levitzki et al.
Trends in pharmacological sciences, 12(5), 171-174 (1991-05-01)
Yan-Jun Xu et al.
Journal of cellular and molecular medicine, 12(3), 942-954 (2008-05-23)
Although lysophosphatidic acid (LPA) is known to increase intracellularfree calcium concentration ([Ca(2+)](i)) in different cell types, the effect of LPA on the skeletal muscle cells is not known. The present study was therefore undertaken to examine the effect of LPA
G E Bilder et al.
The American journal of physiology, 260(4 Pt 1), C721-C730 (1991-04-01)
Tyrphostins are low-molecular-weight synthetic inhibitors of protein tyrosine kinase, which block cell proliferation. Since platelet-derived growth factor (PDGF) is thought to figure prominently in disorders of vascular smooth muscle cells (VSMC), such as atherosclerosis, hypertension, and restenosis, we examined whether
M Zhao et al.
Leukemia, 14(3), 374-378 (2000-03-17)
Somatic mutation of the FLT3 gene, in which the juxtamembrane domain has an internal tandem duplication, is found in 20% of human acute myeloid leukemias and causes constitutive tyrosine phosphorylation of the products. In this study, we observed that the
Manuel Fischer et al.
Molecular biology of the cell, 24(14), 2160-2170 (2013-05-17)
Oxidation of cysteine residues to disulfides drives import of many proteins into the intermembrane space of mitochondria. Recent studies in yeast unraveled the basic principles of mitochondrial protein oxidation, but the kinetics under physiological conditions is unknown. We developed assays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service