Skip to Content
Merck
All Photos(1)

Key Documents

T1694

Sigma-Aldrich

(E)-4-Amino-2-butenoic acid

Synonym(s):

trans-4-Aminocrotonic acid, TACA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO2
CAS Number:
Molecular Weight:
101.10
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

form

powder

color

white

solubility

H2O: 10 mg/mL, clear

SMILES string

NC\C=C\C(O)=O

InChI

1S/C4H7NO2/c5-3-1-2-4(6)7/h1-2H,3,5H2,(H,6,7)/b2-1+

InChI key

FMKJUUQOYOHLTF-OWOJBTEDSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Lu et al.
American journal of physiology. Renal physiology, 308(9), F1020-F1025 (2015-01-23)
Adenosine plays an important role in regulation of renal microcirculation. All receptors of adenosine, A1, A2A, A2B, and A3, have been found in the kidney. However, little is known about the location and function of the A3 receptor in the
D Zhang et al.
Trends in pharmacological sciences, 22(3), 121-132 (2001-03-10)
In less than a decade our knowledge of the GABA(C) receptor, a new type of Cl(-)-permeable ionotropic GABA receptor, has greatly increased based on studies of both native and recombinant receptors. Careful comparison of properties of native and recombinant receptors
Kinga K Borowicz et al.
Pharmacological reports : PR, 57(1), 121-123 (2005-04-26)
In the present study, we evaluated TACA (a potent agonist of GABA(A) and GABA(C) receptors) in the electroconvulsive threshold test in mice. Surprisingly, TACA (at 15 and 25 mg/kg) significantly decreased the threshold. The highest ineffective dose of TACA was
Leonie M Kamminga et al.
PLoS genetics, 8(7), e1002702-e1002702 (2012-07-26)
RNA interference (RNAi)-related pathways affect gene activity by sequence-specific recruitment of Ago proteins to mRNA target molecules. The sequence specificity of this process stems from small RNA (sRNA) co-factors bound by the Ago protein. Stability of sRNA molecules in some
Seung Yun Yang et al.
Nature communications, 4, 1702-1702 (2013-04-18)
Achieving significant adhesion to soft tissues while minimizing tissue damage poses a considerable clinical challenge. Chemical-based adhesives require tissue-specific reactive chemistry, typically inducing a significant inflammatory response. Staples are fraught with limitations including high-localized tissue stress and increased risk of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service