Skip to Content
Merck
All Photos(5)

Key Documents

T1147

Sigma-Aldrich

apo-Transferrin human

powder, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)

Synonym(s):

Human transferrin, Siderophilin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

biological source

human plasma

product line

BioReagent

Assay

≥98% (agarose gel electrophoresis)

form

powder

mol wt

76-81 kDa

concentration

~25 mM

technique(s)

cell culture | mammalian: suitable

impurities

HIV and HBsAg, source material tested negative
≤0.005% iron

solubility

H2O: 50 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TF(7018)

Looking for similar products? Visit Product Comparison Guide

Application

Apo-Transferrin human can be supplemented with iron or used to bind free iron present in media. It is also used to transport iron into mammalian cells.

Biochem/physiol Actions

Transferrin is the primary iron binding and cell delivery molecule present in serum. Serum-free cell culture systems require a delivery format for iron. Transferrin is the preferential delivery form of iron because cells process transferrin bound iron in a physiologically appropriate way through transferrin receptors on the cell surface. Human apo-transferrin is a high affinity transferrin that can be used with a wide range of cells types. Apo-transferrin can be loaded with iron prior to use or added directly to an iron containing medium.

Other Notes

Non-heme iron-transport protein.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giang D Nguyen et al.
PloS one, 8(8), e72698-e72698 (2013-08-24)
Huntington's disease (HD) is a neurodegenerative disease caused by abnormal polyglutamine expansion in the huntingtin protein (Htt). Although both Htt and the HD pathogenic mutation (mHtt) are implicated in early developmental events, their individual involvement has not been adequately explored.
Giang D Nguyen et al.
PloS one, 9(5), e96858-e96858 (2014-05-08)
H1 linker histone proteins are essential for the structural and functional integrity of chromatin and for the fidelity of additional epigenetic modifications. Deletion of H1c, H1d and H1e in mice leads to embryonic lethality by mid-gestation with a broad spectrum
Kathleen N O'Guin et al.
ASN neuro, 6(1), e00135-e00135 (2014-01-31)
The molecular requirements for human myelination are incompletely defined, and further study is needed to fully understand the cellular mechanisms involved during development and in demyelinating diseases. We have established a human co-culture model to study myelination. Our earlier observations
William J Buchser et al.
PloS one, 7(6), e38101-e38101 (2012-06-16)
Trauma to the spinal cord and brain can result in irreparable loss of function. This failure of recovery is in part due to inhibition of axon regeneration by myelin and chondroitin sulfate proteoglycans (CSPGs). Peripheral nervous system (PNS) neurons exhibit
Ayumu Suzuki et al.
Nature communications, 7, 11056-11056 (2016-03-31)
Meiosis is a unique process that allows the generation of reproductive cells. It remains largely unknown how meiosis is initiated in germ cells and why non-germline cells do not undergo meiosis. We previously demonstrated that knockdown of Max expression, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service