Skip to Content
Merck
All Photos(1)

Documents

SRP4580

Sigma-Aldrich

BMP-14 (GDF-5/CDMP-1) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

BMP-14/ CDMP-1, Cartilage derived morphogenetic protein-1, GDF-5, bone morphogenetic protein 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

0.5-4 μg ED50

mol wt

~27 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GDF5(8200)

General description

The gene BMP-14 (bone morphogenetic protein 14) is mapped to human chromosome 20q11.22. It belongs to the bone morphogenetic protein family. Recombinant human BMP-14 is a 27kDa homodimeric protein consisting of two 120 amino acid polypeptide chains.

Biochem/physiol Actions

BMP-14 (bone morphogenetic protein 14) is involved in enhancing proliferations of osteoblasts, periosteum cells and connective tissue fibroblasts. It also promotes endochondral bone growth, proper development of limb skeletons and joints, and odontogenesis. It interacts with BMP type II receptor (BMPR-II) and might be involved in bone morphogenesis. Mutations in the BMP-14 gene are associated with brachydactyly type C, intervertebral disc degeneration, acromesomelic dysplasia Grebe type and osteoarthritis.

Physical form

Lyophilized from 10 mM Sodium Citrate, pH 3.5.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute to a concentration of 0.1-1.0 mg/mL in water containing BSA (50 mg BSA per 1 mg of protein). This solution can then be diluted into other aqueous buffers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of growth differentiation factor-5 and bone morphogenetic protein type II receptor in the development of lumbar intervertebral disc degeneration.
Li YF, et al.
International Journal of Clinical and Experimental Pathology, 8, 719-719 (2015)
Novel homozygous sequence variants in the GDF5 gene underlie acromesomelic dysplasia type-grebe in consanguineous families.
Umair M, et al.
Congenital Anomalies, 57, 45-45 (2017)
A comprehensive meta-analysis of association between genetic variants of GDF5 and osteoarthritis of the knee, hip and hand.
Zhang R, et al.
Inflammation Research, 64, 405-405 (2015)
Two novel homozygous missense mutations in the GDF5 gene cause brachydactyly type C.
Al-Qattan MM, et al.
American Journal of Medical Genetics, 167, 1621-1621 (2015)
Xu-wei Luo et al.
Journal of Zhejiang University. Science. B, 17(1), 30-42 (2016-01-08)
To construct a recombinant adenovirus vector-carrying human growth and differentiation factor-5 (GDF-5) gene, investigate the biological effects of adenovirus-mediated GDF-5 (Ad-GDF-5) on extracellular matrix (ECM) expression in human degenerative disc nucleus pulposus (NP) cells, and explore a candidate gene therapy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service