Skip to Content
Merck
All Photos(1)

Key Documents

SRP3322

Sigma-Aldrich

R-SPONDIN-2 human

recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

RSPO2, Roof plate-specific spondin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

24.4 kDa

packaging

pkg of 20 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RSPO2(340419)

General description

Recombinant human R-Spondin-2 (R-Spo2) is a 24.4kDa protein consisting of 212 amino acid residues. R-Spo2 is encoded by the gene mapped to human chromosome 8q23.1. The encoded protein belongs to the R-spondin family of secreted proteins. Members of RSpo family contain two furin-like cysteine-rich domains at N- terminal end and thrombospondin structural domain and a basic charged C- terminal domain.

Biochem/physiol Actions

R-Spondin 2 (R-Spo2) plays a vital role in the activation of canonical Wnt signaling pathway, which is essential for normal morphogenesis of the respiratory tract and limbs. R-Spo2 positively regulates Wnt-dependent mineralization of osteoblasts, myogenic differentiation and hypertrophic myotube formation. R-Spo2 is also involved in skeletal development. Aberration in the expression of RSPO2 causes ossification of the posterior longitudinal ligament of the spine (OPLL), resulted by the altered differentiation of spinal ligament MSCs (mesenchymal stem cells) into chondrocytes. R-Spo2 is associated with various developmental processes, such as myogenesis, craniofacial morphogenesis, keratinocyte proliferation, osteoblast maturation, lung and limb development. R-Spo2 interacts with leucine-rich-repeat-containing G-protein-coupled receptor 5 (Lgr5) and stimulates acetylcholine receptor (AChR) clustering at the neuromuscular junction (NMJ).

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5 + 150 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-0.5 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Secreted factor R-Spondin 2 is involved in refinement of patterning of the mammalian cochlea.
Mulvaney JF
Developmental Dynamics, 242(2), 179-188 (2013)
R-spondin 2 promotes acetylcholine receptor clustering at the neuromuscular junction via Lgr5.
Nakashima H
Scientific Reports, 6:28512 (2016)
R-spondin 2 is required for normal laryngeal-tracheal, lung and limb morphogenesis.
Bell SM
Development, 135(6), 1049-1058 (2008)
R-Spondin family members regulate the Wnt pathway by a common mechanism.
Kim KA
Molecular Biology of the Cell, 19(6), 2588-2596 (2008)
Masahiro Nakajima et al.
American journal of human genetics, 99(1), 202-207 (2016-07-05)
Ossification of the posterior longitudinal ligament of the spine (OPLL) is a common spinal disorder that results from ectopic ossification of the posterior longitudinal ligament and causes intractable myelopathy and radiculopathy. In a previous genome-wide association study (GWAS), we found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service