SRP3312
IL-10 human
Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Synonym(s):
B-TCGF, CSIF, TGIF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
potency
≤2.0 ng/mL
mol wt
18.6 kDa
packaging
pkg of 10 μg
impurities
endotoxin, tested
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL10(3586)
General description
Interleukin-10 (IL-10) is an immunosuppressive cytokine produced by a variety of mammalian cell types including macrophages, monocytes, T cells, B cells and keratinocytes. Human IL-10 active on murine cells, but murine IL-10 is inactive on human cells. The gene encoding this protein is localized on human chromosome 1q31-32. Recombinant human IL-10 is an 18.6 kDa protein of 161 amino acid residues. Interleukin 10 (IL-10) comprises interferon IFN-γ fold and exists as a symmetric homodimer. It shares structural similarity with interferon γ (IFN-γ).
Application
Interleukin 10 (IL-10) human has been used in enzyme-linked immunosorbent assay.
Biochem/physiol Actions
Interleukin-10 (IL-10) is an anti-inflammatory protein that inhibits the expression of proinflammatory cytokines such as IL-1 and tumor necrosis factor-α (TNF-α). Like IL-4, IL-10 enhances humoral immune responses and attenuates cell-mediated immune reactions. It inhibits the production of IFN-γ and antigen-specific proliferation of type-1 T helper cells (Th1). It protects the host tissue from damage during immune responses. The protein has a role in modulating viral immunity. IL-10 enhances cytokine production, natural killer cell proliferation and cytotoxicity in vitro. IL-10 is an immunosuppressive cytokine. It promotes wound repair and functions to resolve inflammation in the central nervous system. IL-10 mediates its signaling function through the interleukin receptors namely, IL-10RA and two IL-10RBs. The expression of IL-10 is regulated by signal transducer and activator of transcription 3 (STAT3) and suppressor of cytokine signaling-3 (SOCS3) ratios. Polymorphisms in the IL-10 gene is implicated in breast cancer.
Physical form
Lyophilized from 20 mM Sodium Phosphate, pH 7.2.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Amanda R Burmeister et al.
Frontiers in cellular neuroscience, 12, 458-458 (2018-12-14)
Resident cells of the central nervous system (CNS) play an important role in detecting insults and initiating protective or sometimes detrimental host immunity. At peripheral sites, immune responses follow a biphasic course with the rapid, but transient, production of inflammatory
Elnaz Sheikhpour et al.
Reports of biochemistry & molecular biology, 7(1), 30-37 (2018-10-17)
Interleukin (IL)-10, a multifunctional immune-regulatory cytokine with both immunosuppressive and anti-angiogenic functions, is produced by immune cells including macrophages, T lymphocytes, and natural killer cells. Among other effects, IL-10 promotes tumor cell proliferation and metastasis via immunosuppression. Interleukin-10-mediated immunosuppression is
IL-10: A Multifunctional Cytokine in Viral Infections
Jose M. Rojas
Journal of immunology research (2017)
Infection of human macrophages and Dendritic Cells with Mycobacterium tuberculosis Induces a Differential Cytokine Gene Expression That Modulates T Cell response
Journal of Immunology (2001)
K Josephson et al.
Immunity, 15(1), 35-46 (2001-08-04)
Interleukin 10 (IL-10) is a dimeric cytokine that plays a central role in suppressing inflammatory responses. These activities are dependent on the interaction of IL-10 with its high-affinity receptor (IL-10R1). This intermediate complex must subsequently recruit the low-affinity IL-10R2 chain
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service