Skip to Content
Merck
All Photos(1)

Key Documents

SRP3246

Sigma-Aldrich

IP-10 from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CXCL10, crg-2, g-Interferon Inducible Protein 10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

8.6 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.2 EU/μg endotoxin, tested

color

off-white to yellow

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

IP-10 is a CXC chemokine that signals through the CXCR3 receptor. IP-10 selectively chemoattracts Th1 lymphocytes and monocytes, and inhibits cytokine-stimulated hematopoietic progenitor cell proliferation. Additionally, it is angiostatic and mitogenic for vascular smooth muscle cells. Recombinant rat IP-10 is an 8.6 kDa protein containing 77 amino acid residues, including the four conserved cysteine residues present in CXC chemokines.

Biochem/physiol Actions

IP-10 is a CXC chemokine that signals through the CXCR3 receptor. Recombinant rat IP-10 is an 8.6 kDa protein containing 77 amino acid residues, including the four conserved cysteine residues present in CXC chemokines.

Sequence

IPLARTVRCT CIDFHEQPLR PRAIGKLEII PASLSCPHVE IIATMKKNNE KRCLNPESEA IKSLLKAVSQ RRSKRAP

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malefyt, R.
Cytokine Reference, 1061-1037 (2001)
Yingping Hou et al.
The Journal of investigative dermatology, 135(12), 3060-3067 (2015-07-24)
Recessive dystrophic epidermolysis bullosa (RDEB) is an inherited disorder characterized by skin fragility, blistering, and multiple skin wounds with no currently approved or consistently effective treatment. It is due to mutations in the gene encoding type VII collagen (C7). Using
Takanobu Utsumi et al.
The Journal of urology, 192(2), 567-574 (2014-02-13)
Renal cell carcinoma expresses CXCR3 but the function of CXCR3 in renal cell carcinoma has not been clarified. We explored the function of CXCR3 in renal cell carcinoma and investigated CXCR3 regulating factors. We obtained 56 clinical samples of clear cell
P T Mehrotra et al.
Journal of immunology (Baltimore, Md. : 1950), 160(6), 2637-2644 (1998-03-24)
Human NK cell activity can be augmented in vitro by stimulation with IL-2 or IL-12, both of which also induce the production of IFN-gamma, TNF-alpha, and granulocyte-macrophage CSF by NK cells. For the first time, we demonstrate that freshly purified
Ying Wang et al.
PloS one, 9(4), e94696-e94696 (2014-04-16)
Acupuncture is widely used for pain treatment in patients with osteoarthritis or low back pain, but molecular mechanisms remain largely enigmatic. In the early phase of inflammation neutrophilic chemokines direct opioid-containing neutrophils in the inflamed tissue and stimulate opioid peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service