SRP3207
IL-22 from mouse
recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Synonym(s):
IL-TIF
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
biological source
mouse
recombinant
expressed in E. coli
Assay
≥98% (HPLC)
≥98% (SDS-PAGE)
form
lyophilized
mol wt
16.8 kDa
packaging
pkg of 10 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... IL22(50929)
General description
IL-22 is a member of the IL-10 family of regulatory cytokines which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Members of this family share partial homology in their amino acid sequences, but they are dissimilar in their biological functions. Produced by T lymphocytes, IL-22 inhibits IL-4 production by Th2 cells, and induces acute phase reactants in the liver and pancreas. IL-22 signals through a receptor system consisting of IL-10Rb/CRF2-4 and IL-22R, both of which are members of the class II cytokine-receptor family. Recombinant murine IL-22 is a 16.8 kDa non-disulfide-linked monomeric protein containing of two 147 amino acid polypeptide chains.
Biochem/physiol Actions
IL-22 is a member of the IL-10 family of regulatory cytokines which includes IL-10, IL-19, IL-20, IL-22, IL-24 and IL-26. Recombinant murine IL-22 is a 16.8 kDa non-disulfide-linked monomeric protein containing of two 147 amino acid polypeptide chains.
Sequence
MLPVNTRCKL EVSNFQQPYI VNRTFMLAKE ASLADNNTDV RLIGEKLFRG VSAKDQCYLM KQVLNFTLED VLLPQSDRFQ PYMQEVVPFL TKLSNQLSSC HISGDDQNIQ KNVRRLKETV KKLGESGEIK AIGELDLLFM SLRNACV
Physical form
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.5-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sergei V Kotenko
Cytokine & growth factor reviews, 13(3), 223-240 (2002-12-19)
Five novel cytokines (IL-19, IL-20, IL-22 (IL-TIF), IL-24 (human MDA-7, mouse FISP, rat C49A/Mob-5), and IL-26 (AK155)) demonstrating limited primary sequence identity and probable structural homology to IL-10 have been identified. These cellular cytokines, as well as several cytokines encoded
P Conti et al.
Immunology letters, 88(3), 171-174 (2003-08-28)
It has been reported that the CD4+ T cell is a very important source of interleukin 10 (IL-10), while CD8+ cells produce low amounts. IL-10 exerts several immune stimulating, as well as inhibitory effects. There are at least five novel
Miguel A Pineda et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(6), 1492-1503 (2014-02-06)
The parasitic worm-derived immunomodulator ES-62 protects against disease in the mouse collagen-induced arthritis (CIA) model of rheumatoid arthritis (RA) by suppressing pathogenic interleukin-17 (IL-17) responses. The Th17-associated cytokine IL-22 also appears to have a pathogenic role in autoimmune arthritis, particularly
Simona Avitabile et al.
The Journal of investigative dermatology, 135(11), 2862-2870 (2015-07-15)
Impaired re-epithelialization, imbalanced expression of cytokines and growth factors, and vascular disease contribute to healing impairment in diabetes. IL-22, a pro-inflammatory cytokine mediating a cross-talk between immune system and epithelial cells, has been shown to have a role in repair
Dechun Feng et al.
Journal of immunology (Baltimore, Md. : 1950), 193(5), 2512-2518 (2014-07-27)
Acetaminophen (APAP)-induced liver injury (AILI) accounts for half of the acute liver failure cases in the United States. A better understanding of the underlying mechanisms of AILI is necessary for the development of novel antidotes. We found that pretreatment with
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service