Skip to Content
Merck
All Photos(1)

Key Documents

SRP2093

Sigma-Aldrich

SV40 T-antigen from simian virus

recombinant, expressed in insect cells, ≥80% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

biological source

simian virus

recombinant

expressed in insect cells

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~83.3 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

400 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

simian virus ... SV40gp6(1489531)

Biochem/physiol Actions

The large T antigen (TAg) of Simian virus 40 acts as an initiator of DNA replication, participates in cellular transformation, and induces cell growth. A number of functional domains have been characterized including the N-terminal J domain, nuclear localization signal (NLS), internal helicase domain, Rb and topoisomerase I interacting domain, p53 and ATP binding domain. The p53-interacting domain is also essential for binding p300/CBP. Phosphorylation of TAg by several kinases may regulate its function both positively and negatively.
The viral replication of simian virus 40 (SV40) is carried out by a single protein, T-antigen. This feature makes the virus a useful system for understanding the process of replication and to identify the components of replication machinery. It functions as a DNA helicase along with a cellular single-strand DNA binding protein and topoisomerase I or II, leading to extensive origin unwinding and formation of a pre-initiation complex. The T-antigen associates with two tumor suppressor proteins, p53 and the retinoblastoma protein pRb, and inactivates them.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of DNA polymerase alpha-primase with cellular replication protein A and SV40 T antigen.
Dornreiter I
The Embo Journal, 11, 769-776 (1992)
Z Q Pan et al.
The Journal of biological chemistry, 271(36), 22111-22116 (1996-09-06)
The replication of simian virus 40 (SV40) DNA in vitro requires a trimeric single-stranded DNA (ssDNA)-binding protein called HSSB or RPA. HSSB supports the unwinding of DNA containing the SV40 origin in the presence of the viral-encoded T antigen and
Simian virus 40 large T antigen: the puzzle, the pieces, and the emerging picture.
E Fanning
Journal of virology, 66(3), 1289-1293 (1992-03-01)
H H Chao et al.
Molecular and cellular biology, 20(20), 7624-7633 (2000-09-26)
At least three domains of simian virus 40 large T antigen (TAg) participate in cellular transformation. The LXCXE motif of TAg binds to all members of the retinoblastoma protein (pRB) family of tumor suppressors. The N-terminal 70 residues of TAg
Time-sensitive reversal of hyperplasia in transgenic mice expressing SV40 T antigen.
Ewald D
Science, 273, 1384-1386 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service