Skip to Content
Merck
All Photos(1)

Key Documents

SRP2051

Sigma-Aldrich

HMG1, native from calf thymus

≥85% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

bovine (calf) thymus

Assay

≥85% (SDS-PAGE)

form

frozen liquid

mol wt

~24.9 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

900 μg/mL

technique(s)

membrane array: suitable

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Related Categories

Biochem/physiol Actions

High mobility group 1 (HMG1) is a 26 kDa highly conserved non-sequence-specific DNA-binding nuclear protein. Mammalian HMG1 has two homologous DNA-binding domains HMG boxes, A and B (each of 80–90 amino-acid residues), linked by a short basic region to an acidic C-terminal domain containing 30 consecutive Asp and Glu residues. HMG1 has been implicated in a number of fundamental biological processes including transcription, replication and recombination, in which it plays a role in manipulating DNA structure by bending, looping, compaction or unwinding, or by directly contacting with distinct cellular proteins. HMG-1 can act as a repressor, by interacting with TBP to block pre-initiation complex formation or as an activator, by facilitating the binding of various transcription factors to their cognate DNA sequences. Most recently, it was discovered that HMG-1 is a late mediator of delayed endotoxin lethality by activating downstream cytokine release.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cong Pang et al.
Brain sciences, 12(9) (2022-09-24)
Endogenous host-derived molecules named damage-associated molecular patterns (DAMPs) can induce excessive non-sterile inflammatory responses on recognition of specific membrane-tethered receptors. Here in this study, we aimed to explore the role of DAMP molecule HMGB1 in astrocyte-mediated sterile neuroinflammation and the
M E Bianchi et al.
Science (New York, N.Y.), 243(4894 Pt 1), 1056-1059 (1989-02-24)
Cruciform DNA, a non-double helix form of DNA, can be generated as an intermediate in genetic recombination as well as from palindromic sequences under the effect of supercoiling. Eukaryotic cells are equipped with a DNA-binding protein that selectively recognizes cruciform
Structural features of the HMG chromosomal proteins and their genes.
M Bustin et al.
Biochimica et biophysica acta, 1049(3), 231-243 (1990-07-30)
V Zappavigna et al.
The EMBO journal, 15(18), 4981-4991 (1996-09-16)
High mobility group protein 1 (HMG1) is a non-histone, chromatin-associated nuclear protein with a proposed role in the regulation of eukaryotic gene expression. We show that HMG1 interacts with proteins encoded by the HOX gene family by establishing protein-protein contacts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service