Skip to Content
Merck
All Photos(2)

Key Documents

SRP0330

Sigma-Aldrich

BTK active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)

Synonym(s):

AGMX1, BPK, Bruton agammaglobulinemia tyrosine kinase, PSCTK1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

106 kDa

packaging

pkg of 10 μg

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BTK(695)

General description

Human BTK (GenBank Accession No. NM_000061), full length with N-terminal GST-tag, MW= 106 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Liljeroos et al.
Cellular signalling, 19(3), 625-633 (2006-10-06)
Lipoteichoic acid (LTA) of Gram-positive bacteria initiates innate immune responses via Toll-like receptor-2 (TLR2), resulting in the activation of intracellular signaling and production of inflammatory cytokines in macrophages. Although Bruton's tyrosine kinase (Btk) is biologically important molecule implicated in immune
Niklas Feldhahn et al.
The Journal of experimental medicine, 201(11), 1837-1852 (2005-06-09)
Pre-B cells undergo apoptosis unless they are rescued by pre-B cell receptor-dependent survival signals. We previously showed that the BCR-ABL1 kinase that is expressed in pre-B lymphoblastic leukemia bypasses selection for pre-B cell receptor-dependent survival signals. Investigating possible interference of
L L Ong et al.
SLAS discovery : advancing life sciences R & D, 25(2), 176-185 (2019-11-12)
Protein kinases are intensely studied mediators of cellular signaling. While traditional biochemical screens are capable of identifying compounds that modulate kinase activity, these assays are limited in their capability of predicting compound behavior in a cellular environment. Here, we aim
Ximena Zottig et al.
The protein journal, 36(6), 478-488 (2017-10-05)
Recent investigations of Aneurinibacillus thermoaerophilus strains have allowed identification of a unique solvent tolerant lipase, distinct from known lipases. This work reports the expression and purification of this lipase (LipAT) and the first characterization of its structure and temperature and
Miran Rada et al.
Oncotarget, 8(63), 106639-106647 (2018-01-02)
p53 is a tumour suppressor that is activated in response to various types of stress. It is regulated by a complex pattern of over 50 different post-translational modifications, including ubiquitination by the E3 ligase MDM2, which leads to its proteasomal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service